Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160268208-160268469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT80
Entrez Gene ID
57560
Gene Name
intraflagellar transport 80
Gene Aliases
ATD2, SRTD2, WDR56
UniGene
Hs.478095
Gene Chromosome Location
Chr.3: 160256986-160399532 on Build GRCh38
Transcripts
NM_001190241.1 NM_001190242.1 NM_020800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843147 c.2188G>A p.E730K Chr.3: 160268448-160268448 on Build GRCh38
6043722 c.2201G>A p.R734Q Chr.3: 160268435-160268435 on Build GRCh38
6383674 c.2173A>G p.T725A Chr.3: 160268463-160268463 on Build GRCh38
7284338 c.2200C>T p.R734* Chr.3: 160268436-160268436 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769856006 Chr.3: 160268398-160268398 on Build GRCh38
rs1039272217 Chr.3: 160268409-160268409 on Build GRCh38
rs563445545 Chr.3: 160268244-160268244 on Build GRCh38
rs112382107 Chr.3: 160268336-160268336 on Build GRCh38
rs755428490 Chr.3: 160268383-160268383 on Build GRCh38
rs116072970 Chr.3: 160268360-160268360 on Build GRCh38
rs919051681 Chr.3: 160268297-160268297 on Build GRCh38
rs564220393 Chr.3: 160268249-160268249 on Build GRCh38
rs901815416 Chr.3: 160268427-160268427 on Build GRCh38
rs535067699 Chr.3: 160268423-160268423 on Build GRCh38
rs758076802 Chr.3: 160268362-160268362 on Build GRCh38
rs375941259 Chr.3: 160268436-160268436 on Build GRCh38
rs886058129 Chr.3: 160268431-160268431 on Build GRCh38
rs1049253652 Chr.3: 160268372-160268372 on Build GRCh38
rs969072753 Chr.3: 160268438-160268438 on Build GRCh38
rs372855267 Chr.3: 160268377-160268377 on Build GRCh38
rs115849906 Chr.3: 160268393-160268393 on Build GRCh38
rs532099377 Chr.3: 160268271-160268271 on Build GRCh38
rs751433239 Chr.3: 160268406-160268409 on Build GRCh38
rs1003398267 Chr.3: 160268347-160268347 on Build GRCh38
rs780364698 Chr.3: 160268374-160268374 on Build GRCh38
rs372711710 Chr.3: 160268406-160268406 on Build GRCh38
rs992279246 Chr.3: 160268283-160268283 on Build GRCh38

View Full Product Details