Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33085061-33085310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPB1
Entrez Gene ID
3115
Gene Name
major histocompatibility complex, class II, DP beta 1
Gene Aliases
DPB1, HLA-DP, HLA-DP1B, HLA-DPB
UniGene
Hs.485130
Gene Chromosome Location
Chr.6: 33075926-33089696 on Build GRCh38
Transcripts
NM_002121.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211646 c.410T>G p.L137R Chr.6: 33085064-33085064 on Build GRCh38
3928382 c.525C>A p.I175I Chr.6: 33085110-33085110 on Build GRCh38
6106854 c.495G>T p.L165L Chr.6: 33085149-33085149 on Build GRCh38
4406930 c.555T>C p.D185D Chr.6: 33085209-33085209 on Build GRCh38
1443716 c.572delC p.Q193fs*35 Chr.6: 33085157-33085157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042331 Chr.6: 33085173-33085173 on Build GRCh38
rs751221886 Chr.6: 33085094-33085094 on Build GRCh38
rs781427144 Chr.6: 33085277-33085277 on Build GRCh38
rs1071597 Chr.6: 33085209-33085209 on Build GRCh38
rs894270562 Chr.6: 33085128-33085128 on Build GRCh38
rs1042335 Chr.6: 33085181-33085181 on Build GRCh38
rs770371240 Chr.6: 33085174-33085174 on Build GRCh38
rs750357586 Chr.6: 33085116-33085116 on Build GRCh38
rs9277457 Chr.6: 33085249-33085249 on Build GRCh38
rs755847836 Chr.6: 33085231-33085231 on Build GRCh38
rs762588336 Chr.6: 33085185-33085185 on Build GRCh38
rs748284821 Chr.6: 33085149-33085149 on Build GRCh38
rs370864722 Chr.6: 33085200-33085200 on Build GRCh38
rs376906908 Chr.6: 33085137-33085137 on Build GRCh38
rs772135792 Chr.6: 33085158-33085158 on Build GRCh38
rs747228585 Chr.6: 33085259-33085259 on Build GRCh38
rs372905365 Chr.6: 33085089-33085089 on Build GRCh38
rs756011172 Chr.6: 33085118-33085118 on Build GRCh38
rs144181653 Chr.6: 33085171-33085171 on Build GRCh38
rs780393867 Chr.6: 33085157-33085157 on Build GRCh38
rs771222927 Chr.6: 33085168-33085168 on Build GRCh38
rs778011110 Chr.6: 33085159-33085159 on Build GRCh38
rs61736938 Chr.6: 33085142-33085142 on Build GRCh38
rs772872005 Chr.6: 33085182-33085182 on Build GRCh38
rs757047904 Chr.6: 33085111-33085111 on Build GRCh38
rs747234343 Chr.6: 33085162-33085162 on Build GRCh38
rs750173090 Chr.6: 33085222-33085222 on Build GRCh38
rs539695276 Chr.6: 33085248-33085248 on Build GRCh38
rs766369841 Chr.6: 33085233-33085233 on Build GRCh38
rs780989395 Chr.6: 33085112-33085112 on Build GRCh38
rs41559424 Chr.6: 33085221-33085221 on Build GRCh38
rs757437801 Chr.6: 33085273-33085273 on Build GRCh38
rs534162897 Chr.6: 33085090-33085090 on Build GRCh38
rs14362 Chr.6: 33085204-33085204 on Build GRCh38
rs778977608 Chr.6: 33085146-33085146 on Build GRCh38

View Full Product Details