Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139166402-139166641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3829172 c.621C>T p.S207S Chr.6: 139166633-139166633 on Build GRCh38
1754551 c.404A>G p.Y135C Chr.6: 139166416-139166416 on Build GRCh38
1672800 c.521G>A p.R174H Chr.6: 139166533-139166533 on Build GRCh38
4793006 c.623A>T p.E208V Chr.6: 139166635-139166635 on Build GRCh38
3697545 c.447C>A p.G149G Chr.6: 139166459-139166459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747887327 Chr.6: 139166605-139166605 on Build GRCh38
rs780833487 Chr.6: 139166603-139166603 on Build GRCh38
rs139550012 Chr.6: 139166462-139166462 on Build GRCh38
rs1002814746 Chr.6: 139166491-139166491 on Build GRCh38
rs776277277 Chr.6: 139166433-139166433 on Build GRCh38
rs74498924 Chr.6: 139166519-139166519 on Build GRCh38
rs769303187 Chr.6: 139166609-139166609 on Build GRCh38
rs534872191 Chr.6: 139166465-139166465 on Build GRCh38
rs958067181 Chr.6: 139166598-139166598 on Build GRCh38
rs368964706 Chr.6: 139166456-139166456 on Build GRCh38
rs747594392 Chr.6: 139166616-139166618 on Build GRCh38
rs1032503790 Chr.6: 139166526-139166526 on Build GRCh38
rs529104889 Chr.6: 139166594-139166594 on Build GRCh38
rs762507172 Chr.6: 139166610-139166610 on Build GRCh38
rs574457749 Chr.6: 139166477-139166477 on Build GRCh38
rs757396831 Chr.6: 139166479-139166479 on Build GRCh38
rs779904328 Chr.6: 139166555-139166555 on Build GRCh38
rs746780915 Chr.6: 139166565-139166565 on Build GRCh38
rs553001159 Chr.6: 139166468-139166468 on Build GRCh38
rs779002293 Chr.6: 139166486-139166486 on Build GRCh38
rs764862499 Chr.6: 139166439-139166439 on Build GRCh38
rs983188928 Chr.6: 139166567-139166567 on Build GRCh38
rs768560770 Chr.6: 139166569-139166569 on Build GRCh38
rs761042563 Chr.6: 139166467-139166467 on Build GRCh38
rs760164332 Chr.6: 139166460-139166460 on Build GRCh38
rs761364228 Chr.6: 139166438-139166438 on Build GRCh38
rs772410574 Chr.6: 139166453-139166453 on Build GRCh38
rs1013806605 Chr.6: 139166450-139166450 on Build GRCh38
rs758352882 Chr.6: 139166553-139166553 on Build GRCh38

View Full Product Details