Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138555964-138556195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA9
Entrez Gene ID
3313
Gene Name
heat shock protein family A (Hsp70) member 9
Gene Aliases
CRP40, CSA, EVPLS, GRP-75, GRP75, HEL-S-124m, HSPA9B, MOT, MOT2, MTHSP75, PBP74, SAAN, SIDBA4
UniGene
Hs.184233
Gene Chromosome Location
Chr.5: 138554882-138575629 on Build GRCh38
Transcripts
NM_004134.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7272677 c.1976G>A p.R659Q Chr.5: 138556101-138556101 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762960867 Chr.5: 138556013-138556013 on Build GRCh38
rs41296455 Chr.5: 138556168-138556169 on Build GRCh38
rs764021005 Chr.5: 138556015-138556015 on Build GRCh38
rs371887250 Chr.5: 138556152-138556152 on Build GRCh38
rs969505863 Chr.5: 138556087-138556087 on Build GRCh38
rs923414287 Chr.5: 138556132-138556132 on Build GRCh38
rs184962492 Chr.5: 138556175-138556175 on Build GRCh38
rs771270386 Chr.5: 138556101-138556101 on Build GRCh38
rs928003605 Chr.5: 138556102-138556102 on Build GRCh38
rs778180345 Chr.5: 138556089-138556089 on Build GRCh38
rs748706250 Chr.5: 138556075-138556075 on Build GRCh38
rs1027787961 Chr.5: 138556081-138556081 on Build GRCh38
rs375337663 Chr.5: 138556029-138556029 on Build GRCh38
rs1031597405 Chr.5: 138556004-138556004 on Build GRCh38
rs368382143 Chr.5: 138556033-138556033 on Build GRCh38
rs143890160 Chr.5: 138556151-138556151 on Build GRCh38
rs765294292 Chr.5: 138556008-138556008 on Build GRCh38
rs945922593 Chr.5: 138556169-138556169 on Build GRCh38
rs763050434 Chr.5: 138556143-138556143 on Build GRCh38
rs759551229 Chr.5: 138556000-138556000 on Build GRCh38
rs772388447 Chr.5: 138556133-138556134 on Build GRCh38
rs1015173867 Chr.5: 138556003-138556003 on Build GRCh38
rs775601741 Chr.5: 138556009-138556009 on Build GRCh38
rs748287844 Chr.5: 138556050-138556061 on Build GRCh38
rs201871238 Chr.5: 138556026-138556026 on Build GRCh38
rs778107935 Chr.5: 138556060-138556060 on Build GRCh38
rs543954226 Chr.5: 138556083-138556083 on Build GRCh38
rs746063903 Chr.5: 138556104-138556104 on Build GRCh38
rs769786937 Chr.5: 138556119-138556119 on Build GRCh38
rs775679736 Chr.5: 138556121-138556121 on Build GRCh38
rs750023663 Chr.5: 138556036-138556036 on Build GRCh38
rs776724699 Chr.5: 138556103-138556103 on Build GRCh38
rs535687711 Chr.5: 138556155-138556155 on Build GRCh38
rs747364513 Chr.5: 138556094-138556094 on Build GRCh38
rs751393378 Chr.5: 138556020-138556020 on Build GRCh38
rs201515324 Chr.5: 138556044-138556044 on Build GRCh38
rs750061854 Chr.5: 138556164-138556164 on Build GRCh38
rs764108849 Chr.5: 138556149-138556149 on Build GRCh38

View Full Product Details