Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76575532-76575805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001285460.1 NM_006633.3 XM_005248410.2 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163410 c.435G>A p.M145I Chr.5: 76575746-76575746 on Build GRCh38
5403839 c.458G>A p.S153N Chr.5: 76575769-76575769 on Build GRCh38
3828437 c.441T>C p.Y147Y Chr.5: 76575752-76575752 on Build GRCh38
6686911 c.416G>A p.R139Q Chr.5: 76575727-76575727 on Build GRCh38
7372423 c.450C>A p.H150Q Chr.5: 76575761-76575761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755152677 Chr.5: 76575770-76575770 on Build GRCh38
rs759307808 Chr.5: 76575710-76575710 on Build GRCh38
rs768971802 Chr.5: 76575663-76575663 on Build GRCh38
rs900157573 Chr.5: 76575711-76575711 on Build GRCh38
rs761416180 Chr.5: 76575645-76575645 on Build GRCh38
rs557665116 Chr.5: 76575602-76575602 on Build GRCh38
rs979987460 Chr.5: 76575576-76575576 on Build GRCh38
rs367874853 Chr.5: 76575644-76575644 on Build GRCh38
rs773841096 Chr.5: 76575728-76575728 on Build GRCh38
rs767950534 Chr.5: 76575749-76575749 on Build GRCh38
rs1030248501 Chr.5: 76575772-76575772 on Build GRCh38
rs184214697 Chr.5: 76575658-76575658 on Build GRCh38
rs780286309 Chr.5: 76575660-76575660 on Build GRCh38
rs146869045 Chr.5: 76575601-76575601 on Build GRCh38
rs781709647 Chr.5: 76575653-76575653 on Build GRCh38
rs757727131 Chr.5: 76575647-76575647 on Build GRCh38
rs192746554 Chr.5: 76575572-76575572 on Build GRCh38
rs898332905 Chr.5: 76575666-76575666 on Build GRCh38
rs938707379 Chr.5: 76575592-76575592 on Build GRCh38
rs749943435 Chr.5: 76575727-76575727 on Build GRCh38
rs748304303 Chr.5: 76575689-76575689 on Build GRCh38
rs761328324 Chr.5: 76575731-76575731 on Build GRCh38
rs914129409 Chr.5: 76575562-76575562 on Build GRCh38
rs769503917 Chr.5: 76575719-76575719 on Build GRCh38
rs200135774 Chr.5: 76575723-76575723 on Build GRCh38
rs780934953 Chr.5: 76575761-76575761 on Build GRCh38
rs749496418 Chr.5: 76575661-76575661 on Build GRCh38
rs779105840 Chr.5: 76575680-76575680 on Build GRCh38
rs770978497 Chr.5: 76575690-76575690 on Build GRCh38
rs925468676 Chr.5: 76575587-76575587 on Build GRCh38
rs549528805 Chr.5: 76575628-76575628 on Build GRCh38
rs762488312 Chr.5: 76575724-76575724 on Build GRCh38
rs776721097 Chr.5: 76575693-76575693 on Build GRCh38
rs750963939 Chr.5: 76575750-76575750 on Build GRCh38
rs571883802 Chr.5: 76575594-76575594 on Build GRCh38
rs202069494 Chr.5: 76575781-76575781 on Build GRCh38
rs115754709 Chr.5: 76575657-76575657 on Build GRCh38
rs867999465 Chr.5: 76575700-76575700 on Build GRCh38
rs918868074 Chr.5: 76575595-76575595 on Build GRCh38
rs1028066099 Chr.5: 76575732-76575732 on Build GRCh38
rs949406559 Chr.5: 76575607-76575607 on Build GRCh38
rs140724393 Chr.5: 76575751-76575751 on Build GRCh38
rs938064774 Chr.5: 76575592-76575592 on Build GRCh38
rs567909315 Chr.5: 76575631-76575631 on Build GRCh38
rs201291442 Chr.5: 76575762-76575762 on Build GRCh38

View Full Product Details