Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81660479-81660740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPA1
Entrez Gene ID
3612
Gene Name
inositol monophosphatase 1
Gene Aliases
IMP, IMPA
UniGene
Hs.656694
Gene Chromosome Location
Chr.8: 81656916-81686354 on Build GRCh38
Transcripts
NM_001144878.1 NM_001144879.1 NM_005536.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795026 c.698G>A p.G233D Chr.8: 81660536-81660536 on Build GRCh38
4924855 c.632A>G p.Y211C Chr.8: 81660602-81660602 on Build GRCh38
4890760 c.627T>G p.D209E Chr.8: 81660607-81660607 on Build GRCh38
150500 c.572G>A p.R191Q Chr.8: 81660662-81660662 on Build GRCh38
4548842 c.567G>A p.G189G Chr.8: 81660667-81660667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562872543 Chr.8: 81660615-81660615 on Build GRCh38
rs202127208 Chr.8: 81660575-81660575 on Build GRCh38
rs769064564 Chr.8: 81660572-81660572 on Build GRCh38
rs780834689 Chr.8: 81660649-81660649 on Build GRCh38
rs202089380 Chr.8: 81660534-81660534 on Build GRCh38
rs779184992 Chr.8: 81660662-81660662 on Build GRCh38
rs761726155 Chr.8: 81660614-81660614 on Build GRCh38
rs371572810 Chr.8: 81660507-81660507 on Build GRCh38
rs758883028 Chr.8: 81660684-81660684 on Build GRCh38
rs776498442 Chr.8: 81660609-81660609 on Build GRCh38
rs763974663 Chr.8: 81660695-81660695 on Build GRCh38
rs779495553 Chr.8: 81660519-81660519 on Build GRCh38
rs201475007 Chr.8: 81660586-81660586 on Build GRCh38
rs754151177 Chr.8: 81660655-81660655 on Build GRCh38
rs775477038 Chr.8: 81660549-81660549 on Build GRCh38
rs751136547 Chr.8: 81660645-81660645 on Build GRCh38
rs1050474913 Chr.8: 81660535-81660535 on Build GRCh38
rs765899873 Chr.8: 81660634-81660634 on Build GRCh38
rs754560567 Chr.8: 81660646-81660646 on Build GRCh38
rs772816350 Chr.8: 81660506-81660506 on Build GRCh38
rs1009012304 Chr.8: 81660604-81660604 on Build GRCh38
rs200215384 Chr.8: 81660616-81660616 on Build GRCh38
rs775215785 Chr.8: 81660659-81660659 on Build GRCh38
rs778610151 Chr.8: 81660698-81660698 on Build GRCh38
rs200934912 Chr.8: 81660579-81660579 on Build GRCh38
rs762978480 Chr.8: 81660678-81660678 on Build GRCh38
rs149824165 Chr.8: 81660621-81660621 on Build GRCh38
rs760961461 Chr.8: 81660555-81660555 on Build GRCh38
rs780274905 Chr.8: 81660706-81660706 on Build GRCh38
rs200393051 Chr.8: 81660547-81660547 on Build GRCh38

View Full Product Details