Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 2718370-2718595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV2
Entrez Gene ID
169522
Gene Name
potassium voltage-gated channel modifier subfamily V member 2
Gene Aliases
KV11.1, Kv8.2, RCD3B
UniGene
Hs.622675
Gene Chromosome Location
Chr.9: 2717526-2730037 on Build GRCh38
Transcripts
NM_133497.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373287 c.721C>T p.P241S Chr.9: 2718460-2718460 on Build GRCh38
4510616 c.842C>T p.T281I Chr.9: 2718581-2718581 on Build GRCh38
5654814 c.833C>T p.A278V Chr.9: 2718572-2718572 on Build GRCh38
4809185 c.774C>T p.A258A Chr.9: 2718513-2718513 on Build GRCh38
4186616 c.700C>A p.R234S Chr.9: 2718439-2718439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767223506 Chr.9: 2718417-2718417 on Build GRCh38
rs765249083 Chr.9: 2718461-2718461 on Build GRCh38
rs74587818 Chr.9: 2718415-2718415 on Build GRCh38
rs372007584 Chr.9: 2718393-2718393 on Build GRCh38
rs144330480 Chr.9: 2718567-2718567 on Build GRCh38
rs764150150 Chr.9: 2718547-2718547 on Build GRCh38
rs753147277 Chr.9: 2718512-2718512 on Build GRCh38
rs371912406 Chr.9: 2718426-2718426 on Build GRCh38
rs370443689 Chr.9: 2718563-2718563 on Build GRCh38
rs761705988 Chr.9: 2718557-2718557 on Build GRCh38
rs769985957 Chr.9: 2718439-2718439 on Build GRCh38
rs772479912 Chr.9: 2718493-2718493 on Build GRCh38
rs1011858464 Chr.9: 2718553-2718553 on Build GRCh38
rs755632260 Chr.9: 2718420-2718420 on Build GRCh38
rs764550478 Chr.9: 2718414-2718414 on Build GRCh38
rs778059772 Chr.9: 2718525-2718525 on Build GRCh38
rs751450407 Chr.9: 2718516-2718516 on Build GRCh38
rs774250227 Chr.9: 2718548-2718548 on Build GRCh38
rs12237048 Chr.9: 2718534-2718534 on Build GRCh38
rs753904404 Chr.9: 2718520-2718520 on Build GRCh38
rs769345730 Chr.9: 2718543-2718543 on Build GRCh38
rs574636120 Chr.9: 2718453-2718453 on Build GRCh38
rs147022958 Chr.9: 2718501-2718501 on Build GRCh38
rs199923365 Chr.9: 2718566-2718566 on Build GRCh38
rs745887568 Chr.9: 2718434-2718434 on Build GRCh38
rs755071813 Chr.9: 2718521-2718521 on Build GRCh38
rs765931716 Chr.9: 2718422-2718422 on Build GRCh38
rs373932533 Chr.9: 2718546-2718546 on Build GRCh38
rs962948174 Chr.9: 2718562-2718562 on Build GRCh38
rs139027297 Chr.9: 2718517-2718517 on Build GRCh38
rs760295753 Chr.9: 2718497-2718497 on Build GRCh38
rs763616186 Chr.9: 2718411-2718411 on Build GRCh38
rs750673505 Chr.9: 2718472-2718472 on Build GRCh38
rs778833627 Chr.9: 2718395-2718395 on Build GRCh38
rs760387745 Chr.9: 2718457-2718457 on Build GRCh38
rs138231426 Chr.9: 2718544-2718544 on Build GRCh38
rs779485812 Chr.9: 2718488-2718488 on Build GRCh38
rs104894116 Chr.9: 2718506-2718506 on Build GRCh38
rs10967709 Chr.9: 2718498-2718498 on Build GRCh38
rs754107665 Chr.9: 2718559-2718559 on Build GRCh38
rs140144634 Chr.9: 2718572-2718572 on Build GRCh38
rs368923394 Chr.9: 2718481-2718481 on Build GRCh38
rs776522169 Chr.9: 2718403-2718403 on Build GRCh38
rs759416842 Chr.9: 2718458-2718458 on Build GRCh38
rs371445689 Chr.9: 2718442-2718442 on Build GRCh38
rs767476582 Chr.9: 2718558-2718558 on Build GRCh38
rs776156112 Chr.9: 2718410-2718410 on Build GRCh38
rs769005041 Chr.9: 2718483-2718483 on Build GRCh38
rs763922419 Chr.9: 2718513-2718513 on Build GRCh38
rs982964210 Chr.9: 2718438-2718438 on Build GRCh38
rs757378261 Chr.9: 2718429-2718429 on Build GRCh38
rs756448671 Chr.9: 2718473-2718473 on Build GRCh38
rs1011123387 Chr.9: 2718423-2718423 on Build GRCh38
rs755998410 Chr.9: 2718391-2718391 on Build GRCh38
rs563538395 Chr.9: 2718492-2718492 on Build GRCh38
rs149605734 Chr.9: 2718561-2718561 on Build GRCh38
rs1023306854 Chr.9: 2718470-2718470 on Build GRCh38
rs138861317 Chr.9: 2718495-2718495 on Build GRCh38
rs375426599 Chr.9: 2718433-2718433 on Build GRCh38
rs749834841 Chr.9: 2718418-2718418 on Build GRCh38
rs962085824 Chr.9: 2718448-2718448 on Build GRCh38
rs973199066 Chr.9: 2718454-2718454 on Build GRCh38
rs777741747 Chr.9: 2718456-2718456 on Build GRCh38
rs727503975 Chr.9: 2718460-2718460 on Build GRCh38
rs369967074 Chr.9: 2718440-2718440 on Build GRCh38
rs143138491 Chr.9: 2718466-2718466 on Build GRCh38
rs758877052 Chr.9: 2718524-2718524 on Build GRCh38
rs865812782 Chr.9: 2718491-2718491 on Build GRCh38
rs746837965 Chr.9: 2718405-2718405 on Build GRCh38
rs765328422 Chr.9: 2718501-2718503 on Build GRCh38
rs754945632 Chr.9: 2718425-2718425 on Build GRCh38
rs778924998 Chr.9: 2718522-2718522 on Build GRCh38
rs770502498 Chr.9: 2718531-2718531 on Build GRCh38
rs752364913 Chr.9: 2718462-2718462 on Build GRCh38
rs765337150 Chr.9: 2718565-2718565 on Build GRCh38
rs752669658 Chr.9: 2718508-2718508 on Build GRCh38
rs947756576 Chr.9: 2718459-2718459 on Build GRCh38
rs745567670 Chr.9: 2718535-2718535 on Build GRCh38
rs896414258 Chr.9: 2718552-2718552 on Build GRCh38
rs370332122 Chr.9: 2718409-2718409 on Build GRCh38
rs375765488 Chr.9: 2718445-2718445 on Build GRCh38
rs754754580 Chr.9: 2718394-2718394 on Build GRCh38
rs747401501 Chr.9: 2718526-2718526 on Build GRCh38
rs745988476 Chr.9: 2718404-2718404 on Build GRCh38
rs899801446 Chr.9: 2718503-2718503 on Build GRCh38
rs774907955 Chr.9: 2718545-2718545 on Build GRCh38
rs748659440 Chr.9: 2718490-2718490 on Build GRCh38
rs543649807 Chr.9: 2718469-2718469 on Build GRCh38
rs762870229 Chr.9: 2718511-2718511 on Build GRCh38
rs373963150 Chr.9: 2718400-2718400 on Build GRCh38
rs780258945 Chr.9: 2718475-2718475 on Build GRCh38
rs143012910 Chr.9: 2718523-2718523 on Build GRCh38
rs753769991 Chr.9: 2718424-2718424 on Build GRCh38
rs759818370 Chr.9: 2718403-2718408 on Build GRCh38
rs572091494 Chr.9: 2718390-2718390 on Build GRCh38
rs747313664 Chr.9: 2718571-2718571 on Build GRCh38

View Full Product Details