Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122854742-122854997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1R
Entrez Gene ID
9026
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
HIP12, HIP3, ILWEQ
UniGene
Hs.524815
Gene Chromosome Location
Chr.12: 122834498-122862961 on Build GRCh38
Transcripts
NM_001303097.1 NM_001303099.1 NM_003959.2 XM_011538963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4039683 c.762C>T p.H254H Chr.12: 122854948-122854948 on Build GRCh38
4941602 c.728C>T p.A243V Chr.12: 122854914-122854914 on Build GRCh38
4039682 c.755G>A p.R252Q Chr.12: 122854941-122854941 on Build GRCh38
4446189 c.719-3C>T p.? Chr.12: 122854902-122854902 on Build GRCh38
6296846 c.768G>T p.Q256H Chr.12: 122854954-122854954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200749045 Chr.12: 122854973-122854973 on Build GRCh38
rs137994880 Chr.12: 122854948-122854948 on Build GRCh38
rs773105255 Chr.12: 122854858-122854859 on Build GRCh38
rs577913294 Chr.12: 122854769-122854769 on Build GRCh38
rs555083819 Chr.12: 122854951-122854951 on Build GRCh38
rs200151301 Chr.12: 122854929-122854929 on Build GRCh38
rs778307240 Chr.12: 122854899-122854899 on Build GRCh38
rs747454127 Chr.12: 122854976-122854976 on Build GRCh38
rs766521086 Chr.12: 122854942-122854942 on Build GRCh38
rs772612785 Chr.12: 122854891-122854891 on Build GRCh38
rs760702351 Chr.12: 122854941-122854941 on Build GRCh38
rs776125096 Chr.12: 122854768-122854768 on Build GRCh38
rs2292136 Chr.12: 122854797-122854797 on Build GRCh38
rs201854799 Chr.12: 122854862-122854862 on Build GRCh38
rs200700898 Chr.12: 122854949-122854949 on Build GRCh38
rs145314622 Chr.12: 122854940-122854940 on Build GRCh38
rs551339949 Chr.12: 122854915-122854915 on Build GRCh38
rs1021869835 Chr.12: 122854775-122854775 on Build GRCh38
rs186971602 Chr.12: 122854895-122854895 on Build GRCh38
rs758754207 Chr.12: 122854970-122854970 on Build GRCh38
rs1055229997 Chr.12: 122854763-122854763 on Build GRCh38
rs745375370 Chr.12: 122854908-122854908 on Build GRCh38
rs750519866 Chr.12: 122854854-122854854 on Build GRCh38
rs191719039 Chr.12: 122854914-122854914 on Build GRCh38
rs960169069 Chr.12: 122854802-122854802 on Build GRCh38
rs369853543 Chr.12: 122854886-122854886 on Build GRCh38
rs200663785 Chr.12: 122854873-122854873 on Build GRCh38
rs564656610 Chr.12: 122854796-122854796 on Build GRCh38
rs1042961424 Chr.12: 122854780-122854780 on Build GRCh38
rs905999222 Chr.12: 122854776-122854776 on Build GRCh38
rs761560386 Chr.12: 122854936-122854936 on Build GRCh38
rs561852992 Chr.12: 122854830-122854830 on Build GRCh38
rs779300695 Chr.12: 122854887-122854887 on Build GRCh38
rs372393939 Chr.12: 122854850-122854850 on Build GRCh38
rs184666892 Chr.12: 122854838-122854838 on Build GRCh38
rs748504234 Chr.12: 122854889-122854889 on Build GRCh38
rs762846828 Chr.12: 122854880-122854881 on Build GRCh38
rs768224131 Chr.12: 122854920-122854920 on Build GRCh38
rs753089024 Chr.12: 122854962-122854962 on Build GRCh38
rs2292137 Chr.12: 122854811-122854811 on Build GRCh38
rs769222824 Chr.12: 122854913-122854913 on Build GRCh38

View Full Product Details