Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2821592-2821835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITFG2
Entrez Gene ID
55846
Gene Name
integrin alpha FG-GAP repeat containing 2
Gene Aliases
MDS028
UniGene
Hs.446098
Gene Chromosome Location
Chr.12: 2812621-2832217 on Build GRCh38
Transcripts
NM_018463.3 NR_130744.1 XM_005253707.4 XM_005253709.4 XM_017019641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6687685 c.858G>A p.K286K Chr.12: 2821702-2821702 on Build GRCh38
6687686 c.522G>A p.K174K Chr.12: 2821702-2821702 on Build GRCh38
1322565 c.916C>G p.Q306E Chr.12: 2821760-2821760 on Build GRCh38
6448334 c.948+1G>A p.? Chr.12: 2821793-2821793 on Build GRCh38
6136384 c.945C>T p.V315V Chr.12: 2821789-2821789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764117956 Chr.12: 2821628-2821628 on Build GRCh38
rs761394088 Chr.12: 2821649-2821649 on Build GRCh38
rs138970255 Chr.12: 2821792-2821792 on Build GRCh38
rs754241888 Chr.12: 2821758-2821758 on Build GRCh38
rs542578781 Chr.12: 2821672-2821672 on Build GRCh38
rs772817524 Chr.12: 2821797-2821798 on Build GRCh38
rs372965638 Chr.12: 2821614-2821614 on Build GRCh38
rs199819746 Chr.12: 2821691-2821691 on Build GRCh38
rs755881424 Chr.12: 2821793-2821793 on Build GRCh38
rs763035729 Chr.12: 2821810-2821810 on Build GRCh38
rs769394605 Chr.12: 2821772-2821773 on Build GRCh38
rs745966570 Chr.12: 2821641-2821641 on Build GRCh38
rs768326240 Chr.12: 2821667-2821667 on Build GRCh38
rs749754153 Chr.12: 2821626-2821626 on Build GRCh38
rs758447342 Chr.12: 2821771-2821771 on Build GRCh38
rs780510563 Chr.12: 2821621-2821621 on Build GRCh38
rs756517948 Chr.12: 2821624-2821624 on Build GRCh38
rs757652694 Chr.12: 2821759-2821759 on Build GRCh38
rs747385545 Chr.12: 2821648-2821648 on Build GRCh38
rs772548377 Chr.12: 2821706-2821706 on Build GRCh38
rs377499279 Chr.12: 2821811-2821811 on Build GRCh38
rs775655419 Chr.12: 2821652-2821652 on Build GRCh38
rs773254613 Chr.12: 2821795-2821795 on Build GRCh38
rs750966263 Chr.12: 2821764-2821764 on Build GRCh38
rs576647993 Chr.12: 2821770-2821770 on Build GRCh38
rs901206472 Chr.12: 2821785-2821785 on Build GRCh38
rs780796616 Chr.12: 2821787-2821787 on Build GRCh38
rs1018815293 Chr.12: 2821803-2821803 on Build GRCh38
rs766028463 Chr.12: 2821716-2821716 on Build GRCh38
rs779501908 Chr.12: 2821762-2821762 on Build GRCh38
rs998206178 Chr.12: 2821661-2821661 on Build GRCh38
rs61916646 Chr.12: 2821686-2821686 on Build GRCh38
rs558311018 Chr.12: 2821687-2821687 on Build GRCh38
rs1039777486 Chr.12: 2821723-2821723 on Build GRCh38
rs1010456704 Chr.12: 2821730-2821730 on Build GRCh38
rs747818626 Chr.12: 2821628-2821628 on Build GRCh38
rs751242208 Chr.12: 2821739-2821739 on Build GRCh38
rs762584012 Chr.12: 2821712-2821712 on Build GRCh38
rs764298333 Chr.12: 2821754-2821754 on Build GRCh38
rs768931105 Chr.12: 2821778-2821778 on Build GRCh38
rs370620414 Chr.12: 2821632-2821632 on Build GRCh38
rs374043687 Chr.12: 2821814-2821814 on Build GRCh38
rs539694485 Chr.12: 2821677-2821677 on Build GRCh38
rs780169218 Chr.12: 2821777-2821777 on Build GRCh38
rs759327576 Chr.12: 2821745-2821745 on Build GRCh38
rs376681988 Chr.12: 2821667-2821672 on Build GRCh38
rs968714329 Chr.12: 2821784-2821784 on Build GRCh38
rs778257310 Chr.12: 2821625-2821625 on Build GRCh38
rs747276941 Chr.12: 2821656-2821656 on Build GRCh38
rs753071476 Chr.12: 2821617-2821617 on Build GRCh38

View Full Product Details