Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54033410-54033637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC4
Entrez Gene ID
3221
Gene Name
homeobox C4
Gene Aliases
HOX3, HOX3E, cp19
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54056030 on Build GRCh38
Transcripts
NM_014620.5 NM_018953.3 NR_003084.2

Gene Symbol
HOXC5
Entrez Gene ID
3222
Gene Name
homeobox C5
Gene Aliases
CP11, HOX3, HOX3D
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54035361 on Build GRCh38
Transcripts
NM_014620.5 NM_018953.3 NR_003084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
694035 c.397C>A p.Q133K Chr.12: 54033519-54033519 on Build GRCh38
5483777 c.369G>A p.A123A Chr.12: 54033491-54033491 on Build GRCh38
6418655 c.304A>G p.K102E Chr.12: 54033426-54033426 on Build GRCh38
3462753 c.393G>A p.L131L Chr.12: 54033515-54033515 on Build GRCh38
3462752 c.361G>T p.E121* Chr.12: 54033483-54033483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761555118 Chr.12: 54033462-54033462 on Build GRCh38
rs745951201 Chr.12: 54033439-54033439 on Build GRCh38
rs763926170 Chr.12: 54033453-54033453 on Build GRCh38
rs201107649 Chr.12: 54033528-54033528 on Build GRCh38
rs765853011 Chr.12: 54033588-54033588 on Build GRCh38
rs1001521503 Chr.12: 54033461-54033461 on Build GRCh38
rs202063227 Chr.12: 54033576-54033576 on Build GRCh38
rs771438592 Chr.12: 54033480-54033485 on Build GRCh38
rs753354430 Chr.12: 54033590-54033590 on Build GRCh38
rs755580730 Chr.12: 54033483-54033483 on Build GRCh38
rs753488792 Chr.12: 54033490-54033490 on Build GRCh38
rs1027941572 Chr.12: 54033612-54033612 on Build GRCh38
rs757641660 Chr.12: 54033514-54033514 on Build GRCh38
rs140687506 Chr.12: 54033496-54033496 on Build GRCh38
rs767230924 Chr.12: 54033575-54033575 on Build GRCh38
rs769823676 Chr.12: 54033527-54033527 on Build GRCh38
rs768897638 Chr.12: 54033548-54033548 on Build GRCh38
rs994655859 Chr.12: 54033585-54033585 on Build GRCh38
rs754651283 Chr.12: 54033492-54033492 on Build GRCh38
rs777232589 Chr.12: 54033433-54033433 on Build GRCh38
rs757766377 Chr.12: 54033609-54033609 on Build GRCh38
rs750315927 Chr.12: 54033480-54033480 on Build GRCh38
rs761608399 Chr.12: 54033574-54033574 on Build GRCh38
rs373127865 Chr.12: 54033430-54033430 on Build GRCh38
rs200190925 Chr.12: 54033463-54033463 on Build GRCh38
rs370139804 Chr.12: 54033618-54033618 on Build GRCh38
rs746383607 Chr.12: 54033526-54033526 on Build GRCh38
rs774506573 Chr.12: 54033567-54033567 on Build GRCh38
rs781557948 Chr.12: 54033525-54033525 on Build GRCh38
rs779843113 Chr.12: 54033589-54033590 on Build GRCh38
rs747403149 Chr.12: 54033513-54033513 on Build GRCh38
rs754493955 Chr.12: 54033593-54033593 on Build GRCh38
rs1017717478 Chr.12: 54033530-54033530 on Build GRCh38
rs774101949 Chr.12: 54033454-54033454 on Build GRCh38
rs760645865 Chr.12: 54033584-54033584 on Build GRCh38
rs765905719 Chr.12: 54033488-54033488 on Build GRCh38
rs376224435 Chr.12: 54033440-54033440 on Build GRCh38
rs764998400 Chr.12: 54033601-54033601 on Build GRCh38
rs749552740 Chr.12: 54033546-54033546 on Build GRCh38
rs775617536 Chr.12: 54033542-54033542 on Build GRCh38
rs375633977 Chr.12: 54033602-54033602 on Build GRCh38

View Full Product Details