Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18063795-18064066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
UniGene
Hs.567294
Gene Chromosome Location
Chr.19: 18058994-18099027 on Build GRCh38
Transcripts
NM_001290023.1 NM_001290024.1 NM_005535.2 XM_006722741.3 XM_011527966.2 XM_011527967.2 XM_011527968.2 XM_011527969.2 XM_011527970.2 XM_011527971.2 XM_011527972.2 XM_011527973.2 XM_011527974.2 XM_011527975.2 XM_011527976.2 XM_017026762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992987 c.1561C>T p.R521* Chr.19: 18063933-18063933 on Build GRCh38
3099413 c.1550C>T p.T517M Chr.19: 18063944-18063944 on Build GRCh38
3099416 c.1504G>C p.E502Q Chr.19: 18063990-18063990 on Build GRCh38
3099415 c.1522A>G p.S508G Chr.19: 18063972-18063972 on Build GRCh38
3099417 c.1484A>G p.E495G Chr.19: 18064010-18064010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139619268 Chr.19: 18063925-18063925 on Build GRCh38
rs545789499 Chr.19: 18064030-18064030 on Build GRCh38
rs993655665 Chr.19: 18063938-18063938 on Build GRCh38
rs930769046 Chr.19: 18063814-18063814 on Build GRCh38
rs776498539 Chr.19: 18064044-18064044 on Build GRCh38
rs771540567 Chr.19: 18063900-18063900 on Build GRCh38
rs773049771 Chr.19: 18064010-18064010 on Build GRCh38
rs980653981 Chr.19: 18063941-18063941 on Build GRCh38
rs370204086 Chr.19: 18063835-18063835 on Build GRCh38
rs767499649 Chr.19: 18063861-18063861 on Build GRCh38
rs754629779 Chr.19: 18064043-18064044 on Build GRCh38
rs758908533 Chr.19: 18063962-18063962 on Build GRCh38
rs138796805 Chr.19: 18063824-18063824 on Build GRCh38
rs753471142 Chr.19: 18063884-18063884 on Build GRCh38
rs775513511 Chr.19: 18063920-18063920 on Build GRCh38
rs745969950 Chr.19: 18064000-18064000 on Build GRCh38
rs749510884 Chr.19: 18064004-18064004 on Build GRCh38
rs374771268 Chr.19: 18064002-18064002 on Build GRCh38
rs11575935 Chr.19: 18063921-18063921 on Build GRCh38
rs959907651 Chr.19: 18064029-18064029 on Build GRCh38
rs11575932 Chr.19: 18063894-18063894 on Build GRCh38
rs371186623 Chr.19: 18063972-18063972 on Build GRCh38
rs369744899 Chr.19: 18064028-18064028 on Build GRCh38
rs759885958 Chr.19: 18063838-18063838 on Build GRCh38
rs774268416 Chr.19: 18063851-18063851 on Build GRCh38
rs770860296 Chr.19: 18064041-18064041 on Build GRCh38
rs771721483 Chr.19: 18063937-18063937 on Build GRCh38
rs574486067 Chr.19: 18063865-18063865 on Build GRCh38
rs907931334 Chr.19: 18063984-18063984 on Build GRCh38
rs372578469 Chr.19: 18063944-18063944 on Build GRCh38
rs368321453 Chr.19: 18063939-18063939 on Build GRCh38
rs758633561 Chr.19: 18063889-18063889 on Build GRCh38
rs368015601 Chr.19: 18063967-18063967 on Build GRCh38
rs781736642 Chr.19: 18063999-18063999 on Build GRCh38
rs760657739 Chr.19: 18063943-18063943 on Build GRCh38
rs768902454 Chr.19: 18064006-18064006 on Build GRCh38
rs750308865 Chr.19: 18063864-18063864 on Build GRCh38
rs373972439 Chr.19: 18063963-18063963 on Build GRCh38
rs554300618 Chr.19: 18063831-18063831 on Build GRCh38
rs761443845 Chr.19: 18063860-18063860 on Build GRCh38
rs991981668 Chr.19: 18063933-18063933 on Build GRCh38
rs765363101 Chr.19: 18063844-18063844 on Build GRCh38
rs770060017 Chr.19: 18063913-18063913 on Build GRCh38
rs778329113 Chr.19: 18063973-18063973 on Build GRCh38
rs757905417 Chr.19: 18063980-18063980 on Build GRCh38
rs374732718 Chr.19: 18063887-18063887 on Build GRCh38
rs917838985 Chr.19: 18063936-18063936 on Build GRCh38
rs902093406 Chr.19: 18064024-18064024 on Build GRCh38
rs778430238 Chr.19: 18063877-18063877 on Build GRCh38
rs141968777 Chr.19: 18063910-18063910 on Build GRCh38
rs751957558 Chr.19: 18063975-18063975 on Build GRCh38
rs754721392 Chr.19: 18063876-18063876 on Build GRCh38
rs781478903 Chr.19: 18063906-18063906 on Build GRCh38
rs377695964 Chr.19: 18063888-18063888 on Build GRCh38
rs992631726 Chr.19: 18063825-18063825 on Build GRCh38
rs780351670 Chr.19: 18064003-18064003 on Build GRCh38
rs374870218 Chr.19: 18063871-18063871 on Build GRCh38
rs910368200 Chr.19: 18063901-18063901 on Build GRCh38
rs758823470 Chr.19: 18063969-18063969 on Build GRCh38

View Full Product Details