Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62216094-62216347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRH3
Entrez Gene ID
11255
Gene Name
histamine receptor H3
Gene Aliases
GPCR97, HH3R
UniGene
Hs.251399
Gene Chromosome Location
Chr.20: 62213845-62220267 on Build GRCh38
Transcripts
NM_007232.2 XM_005260266.3 XM_017027623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7097713 c.1014G>T p.K338N Chr.20: 62216330-62216330 on Build GRCh38
6993510 c.1182C>T p.Y394Y Chr.20: 62216162-62216162 on Build GRCh38
6993511 c.1182C>T p.Y394Y Chr.20: 62216162-62216162 on Build GRCh38
6017968 c.1096A>C p.I366L Chr.20: 62216248-62216248 on Build GRCh38
7130214 c.1074G>T p.K358N Chr.20: 62216270-62216270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201840302 Chr.20: 62216198-62216198 on Build GRCh38
rs779498395 Chr.20: 62216326-62216326 on Build GRCh38
rs772081169 Chr.20: 62216261-62216261 on Build GRCh38
rs146277958 Chr.20: 62216197-62216197 on Build GRCh38
rs761910268 Chr.20: 62216125-62216125 on Build GRCh38
rs754327084 Chr.20: 62216190-62216190 on Build GRCh38
rs774575400 Chr.20: 62216130-62216130 on Build GRCh38
rs768385563 Chr.20: 62216117-62216117 on Build GRCh38
rs767802174 Chr.20: 62216162-62216162 on Build GRCh38
rs147003708 Chr.20: 62216251-62216251 on Build GRCh38
rs769883694 Chr.20: 62216235-62216235 on Build GRCh38
rs61729552 Chr.20: 62216179-62216179 on Build GRCh38
rs887995922 Chr.20: 62216144-62216144 on Build GRCh38
rs760862489 Chr.20: 62216274-62216274 on Build GRCh38
rs200593418 Chr.20: 62216305-62216305 on Build GRCh38
rs768495579 Chr.20: 62216203-62216203 on Build GRCh38
rs765759670 Chr.20: 62216309-62216309 on Build GRCh38
rs201198332 Chr.20: 62216136-62216136 on Build GRCh38
rs142859348 Chr.20: 62216207-62216207 on Build GRCh38
rs267606041 Chr.20: 62216161-62216161 on Build GRCh38
rs748030505 Chr.20: 62216219-62216219 on Build GRCh38
rs776680369 Chr.20: 62216124-62216124 on Build GRCh38
rs764429203 Chr.20: 62216185-62216185 on Build GRCh38
rs145146001 Chr.20: 62216260-62216260 on Build GRCh38
rs200924312 Chr.20: 62216202-62216202 on Build GRCh38
rs112357993 Chr.20: 62216194-62216194 on Build GRCh38
rs200831890 Chr.20: 62216288-62216288 on Build GRCh38
rs139063566 Chr.20: 62216201-62216201 on Build GRCh38
rs2067476 Chr.20: 62216129-62216129 on Build GRCh38
rs199552270 Chr.20: 62216289-62216289 on Build GRCh38
rs754210266 Chr.20: 62216299-62216299 on Build GRCh38
rs141729367 Chr.20: 62216312-62216312 on Build GRCh38
rs775772906 Chr.20: 62216267-62216267 on Build GRCh38
rs200178576 Chr.20: 62216180-62216180 on Build GRCh38
rs890686938 Chr.20: 62216290-62216290 on Build GRCh38

View Full Product Details