Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206840903-206841176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL19
Entrez Gene ID
29949
Gene Name
interleukin 19
Gene Aliases
IL-10C, MDA1, NG.1, ZMDA1
UniGene
Hs.661017
Gene Chromosome Location
Chr.1: 206798870-206842981 on Build GRCh38
Transcripts
NM_013371.3 NM_153758.2 XM_011509450.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1560308 c.403A>G p.N135D Chr.1: 206841043-206841043 on Build GRCh38
7100173 c.504G>A p.R168R Chr.1: 206841030-206841030 on Build GRCh38
463845 c.391C>T p.Q131* Chr.1: 206841031-206841031 on Build GRCh38
1560307 c.517A>G p.N173D Chr.1: 206841043-206841043 on Build GRCh38
395819 c.518A>G p.N173S Chr.1: 206841044-206841044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748676816 Chr.1: 206840991-206840991 on Build GRCh38
rs778047591 Chr.1: 206841001-206841001 on Build GRCh38
rs756344661 Chr.1: 206840960-206840960 on Build GRCh38
rs570810974 Chr.1: 206840960-206840960 on Build GRCh38
rs993365391 Chr.1: 206841123-206841123 on Build GRCh38
rs762913961 Chr.1: 206841060-206841060 on Build GRCh38
rs763973930 Chr.1: 206840953-206840953 on Build GRCh38
rs1030169258 Chr.1: 206841124-206841124 on Build GRCh38
rs780932355 Chr.1: 206841004-206841004 on Build GRCh38
rs769329582 Chr.1: 206841034-206841034 on Build GRCh38
rs533618202 Chr.1: 206840950-206840950 on Build GRCh38
rs769479140 Chr.1: 206841119-206841120 on Build GRCh38
rs986294907 Chr.1: 206841148-206841148 on Build GRCh38
rs1016620057 Chr.1: 206841072-206841072 on Build GRCh38
rs547024441 Chr.1: 206840961-206840961 on Build GRCh38
rs774926422 Chr.1: 206841045-206841045 on Build GRCh38
rs2243186 Chr.1: 206840965-206840965 on Build GRCh38
rs2243188 Chr.1: 206841127-206841127 on Build GRCh38
rs368040110 Chr.1: 206841115-206841115 on Build GRCh38
rs758973288 Chr.1: 206840992-206840992 on Build GRCh38
rs202020832 Chr.1: 206841095-206841095 on Build GRCh38
rs1013691817 Chr.1: 206840926-206840926 on Build GRCh38
rs2243187 Chr.1: 206841003-206841003 on Build GRCh38
rs755053867 Chr.1: 206840968-206840968 on Build GRCh38
rs374769203 Chr.1: 206840957-206840957 on Build GRCh38
rs745551200 Chr.1: 206841027-206841027 on Build GRCh38
rs747918008 Chr.1: 206841053-206841053 on Build GRCh38
rs768725548 Chr.1: 206841088-206841088 on Build GRCh38
rs778908860 Chr.1: 206840983-206840983 on Build GRCh38

View Full Product Details