Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63612418-63612643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193775 c.14150T>G p.L4717R Chr.15: 63612501-63612501 on Build GRCh38
6679172 c.14228G>A p.R4743Q Chr.15: 63612423-63612423 on Build GRCh38
4056119 c.14196G>T p.E4732D Chr.15: 63612455-63612455 on Build GRCh38
5427346 c.14186G>A p.G4729E Chr.15: 63612465-63612465 on Build GRCh38
4056117 c.14231A>G p.Y4744C Chr.15: 63612420-63612420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766725063 Chr.15: 63612564-63612564 on Build GRCh38
rs916667629 Chr.15: 63612572-63612572 on Build GRCh38
rs566874445 Chr.15: 63612513-63612513 on Build GRCh38
rs749099790 Chr.15: 63612520-63612520 on Build GRCh38
rs923768922 Chr.15: 63612539-63612539 on Build GRCh38
rs778054905 Chr.15: 63612517-63612517 on Build GRCh38
rs766852363 Chr.15: 63612485-63612485 on Build GRCh38
rs765597904 Chr.15: 63612460-63612460 on Build GRCh38
rs760322735 Chr.15: 63612553-63612553 on Build GRCh38
rs1010415717 Chr.15: 63612500-63612500 on Build GRCh38
rs760269349 Chr.15: 63612458-63612458 on Build GRCh38
rs377460066 Chr.15: 63612596-63612596 on Build GRCh38
rs201217308 Chr.15: 63612594-63612594 on Build GRCh38
rs745761319 Chr.15: 63612535-63612535 on Build GRCh38
rs752889427 Chr.15: 63612585-63612585 on Build GRCh38
rs112382815 Chr.15: 63612565-63612565 on Build GRCh38
rs770741060 Chr.15: 63612529-63612529 on Build GRCh38
rs774191570 Chr.15: 63612530-63612530 on Build GRCh38
rs745822250 Chr.15: 63612599-63612599 on Build GRCh38
rs766979337 Chr.15: 63612453-63612453 on Build GRCh38
rs1056541583 Chr.15: 63612562-63612562 on Build GRCh38
rs774921258 Chr.15: 63612551-63612551 on Build GRCh38
rs994369815 Chr.15: 63612614-63612614 on Build GRCh38
rs778723340 Chr.15: 63612597-63612597 on Build GRCh38
rs775152130 Chr.15: 63612457-63612457 on Build GRCh38
rs759013265 Chr.15: 63612447-63612447 on Build GRCh38
rs574610587 Chr.15: 63612589-63612590 on Build GRCh38
rs959784174 Chr.15: 63612625-63612625 on Build GRCh38
rs536629729 Chr.15: 63612482-63612482 on Build GRCh38
rs201286464 Chr.15: 63612479-63612479 on Build GRCh38
rs771931700 Chr.15: 63612603-63612603 on Build GRCh38
rs781372929 Chr.15: 63612506-63612506 on Build GRCh38
rs770844465 Chr.15: 63612443-63612443 on Build GRCh38
rs777902425 Chr.15: 63612593-63612593 on Build GRCh38
rs149568447 Chr.15: 63612512-63612512 on Build GRCh38
rs377743596 Chr.15: 63612544-63612544 on Build GRCh38
rs767707606 Chr.15: 63612569-63612569 on Build GRCh38
rs756246264 Chr.15: 63612586-63612586 on Build GRCh38
rs774335909 Chr.15: 63612446-63612446 on Build GRCh38
rs755140977 Chr.15: 63612491-63612491 on Build GRCh38
rs776332804 Chr.15: 63612563-63612563 on Build GRCh38
rs763801527 Chr.15: 63612556-63612556 on Build GRCh38
rs752075074 Chr.15: 63612490-63612490 on Build GRCh38

View Full Product Details