Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96595456-96595723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL3
Entrez Gene ID
55683
Gene Name
KAT8 regulatory NSL complex subunit 3
Gene Aliases
KIAA1310, NSL3, Rcd1
UniGene
Hs.516341
Gene Chromosome Location
Chr.2: 96593155-96638379 on Build GRCh38
Transcripts
NM_001115016.2 NR_047653.1 NR_047654.1 NR_047655.1 NR_047656.1 NR_047657.1 NR_047658.1 XM_005263984.1 XM_005263987.1 XM_017004470.1 XM_017004473.1 XM_017004474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4624889 c.2089G>T p.A697S Chr.2: 96595619-96595619 on Build GRCh38
4624888 c.2629G>T p.A877S Chr.2: 96595619-96595619 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003979699 Chr.2: 96595636-96595636 on Build GRCh38
rs769092347 Chr.2: 96595647-96595647 on Build GRCh38
rs771952350 Chr.2: 96595608-96595608 on Build GRCh38
rs749545606 Chr.2: 96595645-96595645 on Build GRCh38
rs915579371 Chr.2: 96595684-96595684 on Build GRCh38
rs899084582 Chr.2: 96595559-96595559 on Build GRCh38
rs373514163 Chr.2: 96595671-96595671 on Build GRCh38
rs773278522 Chr.2: 96595668-96595668 on Build GRCh38
rs748149284 Chr.2: 96595605-96595605 on Build GRCh38
rs777386299 Chr.2: 96595623-96595623 on Build GRCh38
rs767952485 Chr.2: 96595666-96595666 on Build GRCh38
rs754652891 Chr.2: 96595584-96595585 on Build GRCh38
rs533830856 Chr.2: 96595694-96595694 on Build GRCh38
rs368848386 Chr.2: 96595659-96595659 on Build GRCh38
rs552282945 Chr.2: 96595520-96595520 on Build GRCh38
rs780323665 Chr.2: 96595571-96595571 on Build GRCh38
rs906392261 Chr.2: 96595478-96595478 on Build GRCh38
rs756300952 Chr.2: 96595565-96595565 on Build GRCh38
rs537412404 Chr.2: 96595560-96595560 on Build GRCh38
rs776433539 Chr.2: 96595644-96595644 on Build GRCh38
rs753619611 Chr.2: 96595583-96595583 on Build GRCh38
rs754817425 Chr.2: 96595595-96595595 on Build GRCh38
rs766745374 Chr.2: 96595531-96595531 on Build GRCh38
rs774667055 Chr.2: 96595649-96595649 on Build GRCh38
rs565403468 Chr.2: 96595542-96595542 on Build GRCh38
rs554855008 Chr.2: 96595631-96595631 on Build GRCh38
rs1027005292 Chr.2: 96595604-96595604 on Build GRCh38
rs778649273 Chr.2: 96595602-96595602 on Build GRCh38
rs568859141 Chr.2: 96595544-96595544 on Build GRCh38
rs780738120 Chr.2: 96595587-96595589 on Build GRCh38
rs534171866 Chr.2: 96595614-96595614 on Build GRCh38
rs752047870 Chr.2: 96595551-96595551 on Build GRCh38
rs770692222 Chr.2: 96595642-96595642 on Build GRCh38
rs1039000305 Chr.2: 96595514-96595514 on Build GRCh38
rs1020195216 Chr.2: 96595693-96595693 on Build GRCh38

View Full Product Details