Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24183127-24183358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
UniGene
Hs.411865
Gene Chromosome Location
Chr.14: 24180219-24188918 on Build GRCh38
Transcripts
NM_024658.3 NR_051979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2032769 c.2123G>T p.C708F Chr.14: 24183354-24183354 on Build GRCh38
432942 c.2256G>C p.V752V Chr.14: 24183141-24183141 on Build GRCh38
954945 c.2157G>T p.E719D Chr.14: 24183320-24183320 on Build GRCh38
7258094 c.2123G>A p.C708Y Chr.14: 24183354-24183354 on Build GRCh38
5596877 c.2142G>A p.R714R Chr.14: 24183335-24183335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887302263 Chr.14: 24183199-24183199 on Build GRCh38
rs979560971 Chr.14: 24183217-24183217 on Build GRCh38
rs777375498 Chr.14: 24183201-24183201 on Build GRCh38
rs747314959 Chr.14: 24183225-24183225 on Build GRCh38
rs754879325 Chr.14: 24183149-24183149 on Build GRCh38
rs778453206 Chr.14: 24183195-24183195 on Build GRCh38
rs772654867 Chr.14: 24183209-24183209 on Build GRCh38
rs777354518 Chr.14: 24183178-24183178 on Build GRCh38
rs781591755 Chr.14: 24183290-24183290 on Build GRCh38
rs755767211 Chr.14: 24183173-24183173 on Build GRCh38
rs767375699 Chr.14: 24183251-24183251 on Build GRCh38
rs201084822 Chr.14: 24183148-24183148 on Build GRCh38
rs769063524 Chr.14: 24183228-24183228 on Build GRCh38
rs752431105 Chr.14: 24183171-24183171 on Build GRCh38
rs373997560 Chr.14: 24183266-24183266 on Build GRCh38
rs544661588 Chr.14: 24183282-24183282 on Build GRCh38
rs966023914 Chr.14: 24183332-24183332 on Build GRCh38
rs770009334 Chr.14: 24183336-24183336 on Build GRCh38
rs767332861 Chr.14: 24183161-24183161 on Build GRCh38
rs759492726 Chr.14: 24183249-24183249 on Build GRCh38
rs1023641645 Chr.14: 24183181-24183181 on Build GRCh38
rs775364818 Chr.14: 24183325-24183325 on Build GRCh38
rs369934334 Chr.14: 24183180-24183180 on Build GRCh38
rs557348319 Chr.14: 24183234-24183234 on Build GRCh38
rs771871884 Chr.14: 24183233-24183233 on Build GRCh38
rs776014653 Chr.14: 24183218-24183218 on Build GRCh38
rs773235707 Chr.14: 24183247-24183247 on Build GRCh38
rs756763891 Chr.14: 24183187-24183187 on Build GRCh38
rs552521751 Chr.14: 24183277-24183277 on Build GRCh38
rs768951909 Chr.14: 24183289-24183289 on Build GRCh38
rs755959515 Chr.14: 24183257-24183257 on Build GRCh38
rs377656550 Chr.14: 24183316-24183316 on Build GRCh38
rs1047578448 Chr.14: 24183186-24183186 on Build GRCh38
rs773284343 Chr.14: 24183337-24183337 on Build GRCh38
rs944189156 Chr.14: 24183206-24183206 on Build GRCh38
rs572506809 Chr.14: 24183281-24183281 on Build GRCh38
rs780775900 Chr.14: 24183287-24183287 on Build GRCh38
rs746403630 Chr.14: 24183196-24183196 on Build GRCh38
rs563725395 Chr.14: 24183267-24183267 on Build GRCh38
rs752625625 Chr.14: 24183255-24183255 on Build GRCh38

View Full Product Details