Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140658616-140658841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IK
Entrez Gene ID
3550
Gene Name
IK cytokine, down-regulator of HLA II
Gene Aliases
CSA2, RED, RER
UniGene
Hs.421245
Gene Chromosome Location
Chr.5: 140647799-140662480 on Build GRCh38
Transcripts
NM_006083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061737 c.927G>T p.K309N Chr.5: 140658753-140658753 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757382041 Chr.5: 140658781-140658781 on Build GRCh38
rs576248886 Chr.5: 140658722-140658722 on Build GRCh38
rs751266066 Chr.5: 140658732-140658732 on Build GRCh38
rs777652761 Chr.5: 140658774-140658774 on Build GRCh38
rs746208201 Chr.5: 140658805-140658805 on Build GRCh38
rs972723997 Chr.5: 140658783-140658783 on Build GRCh38
rs962738430 Chr.5: 140658770-140658770 on Build GRCh38
rs765557480 Chr.5: 140658771-140658772 on Build GRCh38
rs756421261 Chr.5: 140658818-140658818 on Build GRCh38
rs759456452 Chr.5: 140658750-140658750 on Build GRCh38
rs369181961 Chr.5: 140658703-140658703 on Build GRCh38
rs752746502 Chr.5: 140658768-140658768 on Build GRCh38
rs73271559 Chr.5: 140658642-140658642 on Build GRCh38
rs753956864 Chr.5: 140658779-140658779 on Build GRCh38
rs769246047 Chr.5: 140658695-140658695 on Build GRCh38
rs780287497 Chr.5: 140658700-140658700 on Build GRCh38
rs772494398 Chr.5: 140658807-140658807 on Build GRCh38
rs34282994 Chr.5: 140658755-140658755 on Build GRCh38
rs779216756 Chr.5: 140658794-140658794 on Build GRCh38
rs1026169797 Chr.5: 140658816-140658816 on Build GRCh38
rs762449473 Chr.5: 140658719-140658719 on Build GRCh38
rs780527958 Chr.5: 140658813-140658813 on Build GRCh38
rs756246130 Chr.5: 140658772-140658772 on Build GRCh38
rs1003816323 Chr.5: 140658657-140658657 on Build GRCh38

View Full Product Details