Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24337979-24338208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
UniGene
Hs.657920
Gene Chromosome Location
Chr.1: 24319322-24364482 on Build GRCh38
Transcripts
NM_001195010.1 NM_021180.3 NM_198173.2 NM_198174.2 XM_011541869.1 XM_011541870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123220 c.862G>A p.D288N Chr.1: 24338013-24338013 on Build GRCh38
4030442 c.859G>A p.V287M Chr.1: 24337995-24337995 on Build GRCh38
5716461 c.965T>G p.V322G Chr.1: 24338101-24338101 on Build GRCh38
5984038 c.868G>C p.E290Q Chr.1: 24338019-24338019 on Build GRCh38
5716460 c.950T>G p.V317G Chr.1: 24338101-24338101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756861788 Chr.1: 24338066-24338066 on Build GRCh38
rs551243839 Chr.1: 24338155-24338155 on Build GRCh38
rs958323512 Chr.1: 24338175-24338175 on Build GRCh38
rs137880318 Chr.1: 24338101-24338101 on Build GRCh38
rs904252650 Chr.1: 24338162-24338162 on Build GRCh38
rs888205096 Chr.1: 24338151-24338151 on Build GRCh38
rs148250711 Chr.1: 24338057-24338057 on Build GRCh38
rs369797169 Chr.1: 24338099-24338099 on Build GRCh38
rs1051597583 Chr.1: 24338127-24338127 on Build GRCh38
rs112332866 Chr.1: 24338183-24338183 on Build GRCh38
rs921242467 Chr.1: 24338114-24338114 on Build GRCh38
rs759223377 Chr.1: 24338048-24338048 on Build GRCh38
rs546561450 Chr.1: 24338156-24338156 on Build GRCh38
rs762000715 Chr.1: 24338017-24338017 on Build GRCh38
rs752673677 Chr.1: 24338044-24338044 on Build GRCh38
rs767327739 Chr.1: 24338140-24338140 on Build GRCh38
rs753443097 Chr.1: 24338062-24338062 on Build GRCh38
rs375618831 Chr.1: 24338148-24338148 on Build GRCh38
rs1034112521 Chr.1: 24338168-24338168 on Build GRCh38
rs994048844 Chr.1: 24338136-24338136 on Build GRCh38
rs149165410 Chr.1: 24338030-24338030 on Build GRCh38
rs375107232 Chr.1: 24338112-24338112 on Build GRCh38
rs778662501 Chr.1: 24338068-24338068 on Build GRCh38
rs890382438 Chr.1: 24338149-24338149 on Build GRCh38
rs769875117 Chr.1: 24338126-24338126 on Build GRCh38
rs747060661 Chr.1: 24338106-24338107 on Build GRCh38
rs780280734 Chr.1: 24338086-24338086 on Build GRCh38
rs773354574 Chr.1: 24338129-24338129 on Build GRCh38
rs373710872 Chr.1: 24338100-24338100 on Build GRCh38
rs1024974113 Chr.1: 24338188-24338188 on Build GRCh38
rs999923213 Chr.1: 24338166-24338166 on Build GRCh38
rs141594535 Chr.1: 24338013-24338013 on Build GRCh38
rs761831192 Chr.1: 24338125-24338125 on Build GRCh38
rs745531699 Chr.1: 24338079-24338079 on Build GRCh38
rs770614326 Chr.1: 24338186-24338187 on Build GRCh38
rs767218256 Chr.1: 24338043-24338043 on Build GRCh38
rs997790595 Chr.1: 24338173-24338173 on Build GRCh38
rs750665556 Chr.1: 24338031-24338031 on Build GRCh38
rs983008529 Chr.1: 24338053-24338053 on Build GRCh38
rs759267741 Chr.1: 24338040-24338040 on Build GRCh38
rs763149065 Chr.1: 24338130-24338130 on Build GRCh38
rs772416796 Chr.1: 24338084-24338084 on Build GRCh38
rs886037769 Chr.1: 24338067-24338068 on Build GRCh38

View Full Product Details