Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45098254-45098499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS1
Entrez Gene ID
3787
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
Kv9.1
UniGene
Hs.117780
Gene Chromosome Location
Chr.20: 45091214-45101112 on Build GRCh38
Transcripts
NM_001322799.1 NM_002251.4 XM_017027846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632830 c.471G>A p.L157L Chr.20: 45098301-45098301 on Build GRCh38
4624258 c.384G>A p.E128E Chr.20: 45098388-45098388 on Build GRCh38
117070 c.496G>T p.A166S Chr.20: 45098276-45098276 on Build GRCh38
4620771 c.482T>C p.L161P Chr.20: 45098290-45098290 on Build GRCh38
6855573 c.460G>A p.A154T Chr.20: 45098312-45098312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764547218 Chr.20: 45098338-45098338 on Build GRCh38
rs924554573 Chr.20: 45098456-45098456 on Build GRCh38
rs747093109 Chr.20: 45098457-45098457 on Build GRCh38
rs968994291 Chr.20: 45098408-45098408 on Build GRCh38
rs776149926 Chr.20: 45098313-45098313 on Build GRCh38
rs776913037 Chr.20: 45098342-45098342 on Build GRCh38
rs766016752 Chr.20: 45098365-45098365 on Build GRCh38
rs774683642 Chr.20: 45098286-45098286 on Build GRCh38
rs552417867 Chr.20: 45098345-45098345 on Build GRCh38
rs866211337 Chr.20: 45098449-45098449 on Build GRCh38
rs532420183 Chr.20: 45098337-45098337 on Build GRCh38
rs779334922 Chr.20: 45098436-45098436 on Build GRCh38
rs754498637 Chr.20: 45098379-45098379 on Build GRCh38
rs371977105 Chr.20: 45098281-45098281 on Build GRCh38
rs1002984834 Chr.20: 45098288-45098288 on Build GRCh38
rs770087624 Chr.20: 45098479-45098479 on Build GRCh38
rs554416667 Chr.20: 45098474-45098474 on Build GRCh38
rs749076353 Chr.20: 45098415-45098415 on Build GRCh38
rs762209311 Chr.20: 45098361-45098361 on Build GRCh38
rs993471960 Chr.20: 45098315-45098315 on Build GRCh38
rs776877543 Chr.20: 45098362-45098362 on Build GRCh38
rs746010157 Chr.20: 45098287-45098287 on Build GRCh38
rs985105120 Chr.20: 45098333-45098333 on Build GRCh38
rs751203756 Chr.20: 45098376-45098376 on Build GRCh38
rs761018535 Chr.20: 45098324-45098324 on Build GRCh38
rs755958406 Chr.20: 45098406-45098406 on Build GRCh38
rs772262134 Chr.20: 45098304-45098304 on Build GRCh38
rs978599031 Chr.20: 45098430-45098430 on Build GRCh38
rs1040376123 Chr.20: 45098358-45098358 on Build GRCh38
rs201080873 Chr.20: 45098394-45098394 on Build GRCh38
rs777743004 Chr.20: 45098410-45098410 on Build GRCh38
rs769086547 Chr.20: 45098465-45098465 on Build GRCh38
rs762145037 Chr.20: 45098478-45098478 on Build GRCh38
rs749545248 Chr.20: 45098276-45098276 on Build GRCh38
rs534791107 Chr.20: 45098441-45098441 on Build GRCh38
rs752664327 Chr.20: 45098402-45098402 on Build GRCh38
rs757070162 Chr.20: 45098416-45098416 on Build GRCh38
rs772280462 Chr.20: 45098442-45098442 on Build GRCh38
rs1017677972 Chr.20: 45098404-45098404 on Build GRCh38

View Full Product Details