Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185190065-185190334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP97
Entrez Gene ID
57587
Gene Name
cilia and flagella associated protein 97
Gene Aliases
KIAA1430, hmw
UniGene
Hs.535734
Gene Chromosome Location
Chr.4: 185159662-185210251 on Build GRCh38
Transcripts
NM_001292033.1 NM_020827.2 XM_017008483.1 XM_017008484.1 XM_017008485.1 XM_017008486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1212185 c.956T>A p.V319D Chr.4: 185190241-185190241 on Build GRCh38
3130414 c.932G>A p.G311E Chr.4: 185190265-185190265 on Build GRCh38
7373653 c.886G>A p.E296K Chr.4: 185190311-185190311 on Build GRCh38
3130412 c.994G>A p.D332N Chr.4: 185190203-185190203 on Build GRCh38
7373652 c.886G>A p.E296K Chr.4: 185190311-185190311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76801972 Chr.4: 185190124-185190124 on Build GRCh38
rs776291545 Chr.4: 185190244-185190244 on Build GRCh38
rs750242030 Chr.4: 185190187-185190187 on Build GRCh38
rs766427099 Chr.4: 185190228-185190228 on Build GRCh38
rs371400155 Chr.4: 185190194-185190194 on Build GRCh38
rs763608646 Chr.4: 185190241-185190241 on Build GRCh38
rs532611860 Chr.4: 185190232-185190232 on Build GRCh38
rs756179538 Chr.4: 185190198-185190198 on Build GRCh38
rs758330573 Chr.4: 185190189-185190189 on Build GRCh38
rs145238765 Chr.4: 185190152-185190152 on Build GRCh38
rs371716078 Chr.4: 185190145-185190145 on Build GRCh38
rs1018888538 Chr.4: 185190302-185190302 on Build GRCh38
rs981765763 Chr.4: 185190171-185190171 on Build GRCh38
rs374331121 Chr.4: 185190100-185190100 on Build GRCh38
rs761484859 Chr.4: 185190248-185190248 on Build GRCh38
rs375722943 Chr.4: 185190247-185190247 on Build GRCh38
rs753765509 Chr.4: 185190167-185190167 on Build GRCh38
rs760861278 Chr.4: 185190124-185190126 on Build GRCh38
rs1036704295 Chr.4: 185190245-185190245 on Build GRCh38
rs769007342 Chr.4: 185190098-185190098 on Build GRCh38
rs763483267 Chr.4: 185190117-185190117 on Build GRCh38
rs756988887 Chr.4: 185190210-185190210 on Build GRCh38
rs758236658 Chr.4: 185190260-185190260 on Build GRCh38
rs757385180 Chr.4: 185190294-185190294 on Build GRCh38
rs769813500 Chr.4: 185190103-185190103 on Build GRCh38
rs765024187 Chr.4: 185190231-185190232 on Build GRCh38
rs552444758 Chr.4: 185190253-185190253 on Build GRCh38
rs760206464 Chr.4: 185190155-185190155 on Build GRCh38
rs766118958 Chr.4: 185190268-185190268 on Build GRCh38
rs775673590 Chr.4: 185190101-185190103 on Build GRCh38
rs763548233 Chr.4: 185190160-185190160 on Build GRCh38
rs765059730 Chr.4: 185190183-185190183 on Build GRCh38
rs988827613 Chr.4: 185190137-185190137 on Build GRCh38
rs763917289 Chr.4: 185190127-185190131 on Build GRCh38
rs752917017 Chr.4: 185190276-185190276 on Build GRCh38
rs754102167 Chr.4: 185190252-185190252 on Build GRCh38
rs774708963 Chr.4: 185190139-185190139 on Build GRCh38
rs200841332 Chr.4: 185190229-185190229 on Build GRCh38
rs749740526 Chr.4: 185190200-185190200 on Build GRCh38
rs757280079 Chr.4: 185190175-185190175 on Build GRCh38
rs142184332 Chr.4: 185190193-185190193 on Build GRCh38
rs756304814 Chr.4: 185190277-185190277 on Build GRCh38
rs753854333 Chr.4: 185190185-185190186 on Build GRCh38
rs868245217 Chr.4: 185190265-185190266 on Build GRCh38
rs771430753 Chr.4: 185190203-185190203 on Build GRCh38
rs760690401 Chr.4: 185190172-185190172 on Build GRCh38
rs777738621 Chr.4: 185190199-185190199 on Build GRCh38
rs773738540 Chr.4: 185190115-185190115 on Build GRCh38
rs774904788 Chr.4: 185190222-185190222 on Build GRCh38
rs759120701 Chr.4: 185190140-185190140 on Build GRCh38
rs748249865 Chr.4: 185190102-185190102 on Build GRCh38
rs749364561 Chr.4: 185190279-185190279 on Build GRCh38

View Full Product Details