Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123645388-123645627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6867323 c.120C>A p.A40A Chr.12: 123645481-123645481 on Build GRCh38
692511 c.126G>C p.M42I Chr.12: 123645487-123645487 on Build GRCh38
1628440 c.157C>T p.R53C Chr.12: 123645518-123645518 on Build GRCh38
6853005 c.128T>C p.V43A Chr.12: 123645489-123645489 on Build GRCh38
6676710 c.119C>T p.A40V Chr.12: 123645480-123645480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765550984 Chr.12: 123645439-123645439 on Build GRCh38
rs752205665 Chr.12: 123645543-123645543 on Build GRCh38
rs753398252 Chr.12: 123645443-123645443 on Build GRCh38
rs376533282 Chr.12: 123645482-123645482 on Build GRCh38
rs138518883 Chr.12: 123645512-123645512 on Build GRCh38
rs755065949 Chr.12: 123645523-123645523 on Build GRCh38
rs1016153414 Chr.12: 123645514-123645514 on Build GRCh38
rs1052405177 Chr.12: 123645495-123645495 on Build GRCh38
rs764822420 Chr.12: 123645526-123645526 on Build GRCh38
rs147119955 Chr.12: 123645502-123645502 on Build GRCh38
rs751327361 Chr.12: 123645566-123645566 on Build GRCh38
rs79670068 Chr.12: 123645447-123645447 on Build GRCh38
rs759161115 Chr.12: 123645525-123645525 on Build GRCh38
rs2292702 Chr.12: 123645581-123645581 on Build GRCh38
rs756233509 Chr.12: 123645586-123645586 on Build GRCh38
rs769858898 Chr.12: 123645501-123645501 on Build GRCh38
rs758020647 Chr.12: 123645550-123645550 on Build GRCh38
rs372822888 Chr.12: 123645508-123645508 on Build GRCh38
rs761380347 Chr.12: 123645438-123645439 on Build GRCh38
rs183940144 Chr.12: 123645517-123645517 on Build GRCh38
rs768094576 Chr.12: 123645429-123645429 on Build GRCh38
rs763737496 Chr.12: 123645559-123645559 on Build GRCh38
rs374884792 Chr.12: 123645448-123645448 on Build GRCh38
rs746780189 Chr.12: 123645481-123645481 on Build GRCh38
rs780018165 Chr.12: 123645595-123645595 on Build GRCh38
rs764332887 Chr.12: 123645413-123645413 on Build GRCh38
rs752264755 Chr.12: 123645449-123645449 on Build GRCh38
rs745773495 Chr.12: 123645585-123645585 on Build GRCh38
rs865911942 Chr.12: 123645542-123645542 on Build GRCh38
rs746005568 Chr.12: 123645492-123645492 on Build GRCh38
rs758264498 Chr.12: 123645453-123645453 on Build GRCh38
rs3817308 Chr.12: 123645422-123645422 on Build GRCh38
rs781051083 Chr.12: 123645483-123645483 on Build GRCh38
rs781172338 Chr.12: 123645582-123645582 on Build GRCh38
rs752156600 Chr.12: 123645415-123645415 on Build GRCh38
rs750808915 Chr.12: 123645547-123645549 on Build GRCh38
rs141727968 Chr.12: 123645473-123645473 on Build GRCh38
rs530270065 Chr.12: 123645518-123645518 on Build GRCh38

View Full Product Details