Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45714067-45714292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPP
Entrez Gene ID
3652
Gene Name
intracisternal A particle-promoted polypeptide
Gene Aliases
KLHL27
UniGene
Hs.744252
Gene Chromosome Location
Chr.1: 45694324-45750813 on Build GRCh38
Transcripts
NM_001145349.1 NM_005897.2 XM_005270839.4 XM_006710623.3 XM_011541394.2 XM_017001210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5485705 c.1521C>A p.S507S Chr.1: 45714255-45714255 on Build GRCh38
5485706 c.1521C>A p.S507S Chr.1: 45714255-45714255 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938424378 Chr.1: 45714162-45714162 on Build GRCh38
rs79983587 Chr.1: 45714202-45714202 on Build GRCh38
rs758794945 Chr.1: 45714212-45714212 on Build GRCh38
rs994868614 Chr.1: 45714241-45714241 on Build GRCh38
rs1041544038 Chr.1: 45714141-45714141 on Build GRCh38
rs894115671 Chr.1: 45714176-45714176 on Build GRCh38
rs558740156 Chr.1: 45714134-45714134 on Build GRCh38
rs755236274 Chr.1: 45714251-45714251 on Build GRCh38
rs1005846475 Chr.1: 45714196-45714196 on Build GRCh38
rs1055962753 Chr.1: 45714133-45714134 on Build GRCh38
rs1025792652 Chr.1: 45714245-45714245 on Build GRCh38
rs186262832 Chr.1: 45714103-45714103 on Build GRCh38
rs763456235 Chr.1: 45714208-45714208 on Build GRCh38
rs780421600 Chr.1: 45714240-45714240 on Build GRCh38
rs758385427 Chr.1: 45714146-45714146 on Build GRCh38
rs751903282 Chr.1: 45714246-45714246 on Build GRCh38
rs897513676 Chr.1: 45714189-45714189 on Build GRCh38
rs781384084 Chr.1: 45714252-45714252 on Build GRCh38
rs534097163 Chr.1: 45714130-45714130 on Build GRCh38
rs928494859 Chr.1: 45714145-45714145 on Build GRCh38
rs180806130 Chr.1: 45714195-45714195 on Build GRCh38
rs556628475 Chr.1: 45714199-45714199 on Build GRCh38
rs990042481 Chr.1: 45714111-45714111 on Build GRCh38
rs187985613 Chr.1: 45714164-45714164 on Build GRCh38

View Full Product Details