Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69115059-69115294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAS3
Entrez Gene ID
3038
Gene Name
hyaluronan synthase 3
Gene Aliases
-
UniGene
Hs.592069
Gene Chromosome Location
Chr.16: 69105564-69118719 on Build GRCh38
Transcripts
NM_001199280.1 NM_005329.2 NM_138612.2 XM_005255921.2 XM_011523061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417300 c.1528C>A p.L510I Chr.16: 69115132-69115132 on Build GRCh38
6255101 c.1565G>T p.C522F Chr.16: 69115169-69115169 on Build GRCh38
471998 c.1642T>C p.L548L Chr.16: 69115246-69115246 on Build GRCh38
6485151 c.1536C>T p.F512F Chr.16: 69115140-69115140 on Build GRCh38
6677702 c.1605C>T p.I535I Chr.16: 69115209-69115209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756362947 Chr.16: 69115231-69115231 on Build GRCh38
rs758335498 Chr.16: 69115236-69115236 on Build GRCh38
rs899134890 Chr.16: 69115107-69115107 on Build GRCh38
rs753891032 Chr.16: 69115234-69115234 on Build GRCh38
rs751891527 Chr.16: 69115078-69115078 on Build GRCh38
rs767900238 Chr.16: 69115228-69115228 on Build GRCh38
rs776247498 Chr.16: 69115206-69115206 on Build GRCh38
rs948852773 Chr.16: 69115090-69115090 on Build GRCh38
rs762251434 Chr.16: 69115219-69115219 on Build GRCh38
rs372206673 Chr.16: 69115261-69115261 on Build GRCh38
rs993475369 Chr.16: 69115120-69115120 on Build GRCh38
rs778949811 Chr.16: 69115137-69115137 on Build GRCh38
rs977762488 Chr.16: 69115232-69115232 on Build GRCh38
rs768024807 Chr.16: 69115272-69115272 on Build GRCh38
rs563384214 Chr.16: 69115098-69115098 on Build GRCh38
rs2232233 Chr.16: 69115270-69115270 on Build GRCh38
rs150339274 Chr.16: 69115132-69115132 on Build GRCh38
rs368360996 Chr.16: 69115255-69115255 on Build GRCh38
rs763380500 Chr.16: 69115215-69115215 on Build GRCh38
rs748064844 Chr.16: 69115149-69115149 on Build GRCh38
rs2232232 Chr.16: 69115194-69115194 on Build GRCh38
rs534959741 Chr.16: 69115217-69115217 on Build GRCh38
rs546894403 Chr.16: 69115230-69115230 on Build GRCh38
rs373836879 Chr.16: 69115214-69115214 on Build GRCh38
rs762308758 Chr.16: 69115271-69115271 on Build GRCh38
rs781556727 Chr.16: 69115121-69115121 on Build GRCh38
rs1009011245 Chr.16: 69115144-69115144 on Build GRCh38
rs780928740 Chr.16: 69115253-69115253 on Build GRCh38
rs754089085 Chr.16: 69115136-69115139 on Build GRCh38
rs144185943 Chr.16: 69115180-69115180 on Build GRCh38
rs746856097 Chr.16: 69115242-69115242 on Build GRCh38
rs759090231 Chr.16: 69115209-69115209 on Build GRCh38
rs375357316 Chr.16: 69115246-69115246 on Build GRCh38
rs780018082 Chr.16: 69115233-69115233 on Build GRCh38
rs745999540 Chr.16: 69115126-69115126 on Build GRCh38
rs764761950 Chr.16: 69115213-69115213 on Build GRCh38
rs774865070 Chr.16: 69115259-69115259 on Build GRCh38
rs770925236 Chr.16: 69115267-69115267 on Build GRCh38
rs139637191 Chr.16: 69115155-69115155 on Build GRCh38
rs746803121 Chr.16: 69115185-69115185 on Build GRCh38
rs151020212 Chr.16: 69115254-69115254 on Build GRCh38
rs112187376 Chr.16: 69115181-69115181 on Build GRCh38
rs916444670 Chr.16: 69115257-69115257 on Build GRCh38
rs777753467 Chr.16: 69115239-69115239 on Build GRCh38

View Full Product Details