Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1791126-1791399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFALS
Entrez Gene ID
3483
Gene Name
insulin like growth factor binding protein acid labile subunit
Gene Aliases
ACLSD, ALS
UniGene
Hs.839
Gene Chromosome Location
Chr.16: 1790413-1794908 on Build GRCh38
Transcripts
NM_001146006.1 NM_004970.2 NR_027389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4058752 c.1072T>G p.F358V Chr.16: 1791346-1791346 on Build GRCh38
4058755 c.1134T>C p.L378L Chr.16: 1791398-1791398 on Build GRCh38
6684306 c.1321C>A p.P441T Chr.16: 1791211-1791211 on Build GRCh38
4058756 c.1020T>C p.L340L Chr.16: 1791398-1791398 on Build GRCh38
4691947 c.1381G>A p.V461M Chr.16: 1791151-1791151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759124435 Chr.16: 1791280-1791280 on Build GRCh38
rs777001399 Chr.16: 1791166-1791166 on Build GRCh38
rs758316085 Chr.16: 1791365-1791365 on Build GRCh38
rs145307041 Chr.16: 1791248-1791248 on Build GRCh38
rs760574429 Chr.16: 1791323-1791323 on Build GRCh38
rs761538619 Chr.16: 1791254-1791254 on Build GRCh38
rs755011133 Chr.16: 1791354-1791354 on Build GRCh38
rs748115182 Chr.16: 1791301-1791301 on Build GRCh38
rs772876504 Chr.16: 1791266-1791266 on Build GRCh38
rs1034369449 Chr.16: 1791194-1791194 on Build GRCh38
rs200778437 Chr.16: 1791353-1791353 on Build GRCh38
rs773502836 Chr.16: 1791324-1791324 on Build GRCh38
rs774118306 Chr.16: 1791182-1791182 on Build GRCh38
rs765834680 Chr.16: 1791195-1791195 on Build GRCh38
rs749137957 Chr.16: 1791271-1791271 on Build GRCh38
rs778249405 Chr.16: 1791157-1791157 on Build GRCh38
rs764069429 Chr.16: 1791181-1791181 on Build GRCh38
rs771872163 Chr.16: 1791261-1791261 on Build GRCh38
rs755597815 Chr.16: 1791193-1791193 on Build GRCh38
rs780388342 Chr.16: 1791294-1791294 on Build GRCh38
rs752299042 Chr.16: 1791335-1791335 on Build GRCh38
rs995290554 Chr.16: 1791359-1791359 on Build GRCh38
rs36068954 Chr.16: 1791152-1791152 on Build GRCh38
rs766102791 Chr.16: 1791275-1791275 on Build GRCh38
rs756502511 Chr.16: 1791350-1791350 on Build GRCh38
rs202131276 Chr.16: 1791285-1791285 on Build GRCh38
rs35128702 Chr.16: 1791223-1791223 on Build GRCh38
rs750852124 Chr.16: 1791287-1791287 on Build GRCh38
rs753116894 Chr.16: 1791158-1791158 on Build GRCh38
rs17855510 Chr.16: 1791328-1791328 on Build GRCh38
rs1046208003 Chr.16: 1791377-1791377 on Build GRCh38
rs768131429 Chr.16: 1791343-1791343 on Build GRCh38
rs375344055 Chr.16: 1791341-1791341 on Build GRCh38
rs767089671 Chr.16: 1791189-1791189 on Build GRCh38
rs762769857 Chr.16: 1791180-1791180 on Build GRCh38
rs199573954 Chr.16: 1791148-1791148 on Build GRCh38
rs34139827 Chr.16: 1791203-1791203 on Build GRCh38
rs945016899 Chr.16: 1791372-1791372 on Build GRCh38
rs1005976537 Chr.16: 1791198-1791198 on Build GRCh38
rs777773612 Chr.16: 1791368-1791368 on Build GRCh38
rs753286446 Chr.16: 1791277-1791277 on Build GRCh38
rs758807328 Chr.16: 1791197-1791197 on Build GRCh38
rs147555919 Chr.16: 1791270-1791270 on Build GRCh38
rs779977238 Chr.16: 1791235-1791235 on Build GRCh38
rs769786721 Chr.16: 1791174-1791174 on Build GRCh38
rs775458081 Chr.16: 1791178-1791178 on Build GRCh38
rs776108311 Chr.16: 1791374-1791374 on Build GRCh38
rs753426055 Chr.16: 1791196-1791196 on Build GRCh38
rs886051777 Chr.16: 1791245-1791245 on Build GRCh38
rs772037518 Chr.16: 1791319-1791319 on Build GRCh38
rs764642331 Chr.16: 1791284-1791284 on Build GRCh38
rs760211480 Chr.16: 1791271-1791271 on Build GRCh38
rs149041489 Chr.16: 1791295-1791295 on Build GRCh38
rs370524604 Chr.16: 1791356-1791356 on Build GRCh38
rs781380507 Chr.16: 1791286-1791286 on Build GRCh38
rs771276931 Chr.16: 1791159-1791159 on Build GRCh38
rs200564045 Chr.16: 1791151-1791151 on Build GRCh38
rs778769182 Chr.16: 1791296-1791296 on Build GRCh38
rs770783693 Chr.16: 1791371-1791371 on Build GRCh38
rs112377196 Chr.16: 1791349-1791349 on Build GRCh38
rs748274787 Chr.16: 1791361-1791361 on Build GRCh38
rs761661590 Chr.16: 1791188-1791188 on Build GRCh38
rs370923559 Chr.16: 1791214-1791214 on Build GRCh38
rs781519921 Chr.16: 1791219-1791219 on Build GRCh38
rs756279959 Chr.16: 1791293-1791293 on Build GRCh38
rs771730351 Chr.16: 1791320-1791320 on Build GRCh38
rs764871476 Chr.16: 1791329-1791329 on Build GRCh38
rs750057452 Chr.16: 1791191-1791191 on Build GRCh38
rs768574755 Chr.16: 1791247-1791247 on Build GRCh38
rs867401844 Chr.16: 1791318-1791318 on Build GRCh38
rs778492891 Chr.16: 1791210-1791210 on Build GRCh38
rs745907174 Chr.16: 1791167-1791167 on Build GRCh38
rs746108965 Chr.16: 1791220-1791220 on Build GRCh38
rs749440882 Chr.16: 1791242-1791242 on Build GRCh38
rs370544997 Chr.16: 1791213-1791213 on Build GRCh38

View Full Product Details