Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132136990-132137263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
UniGene
Hs.106254
Gene Chromosome Location
Chr.10: 132036068-132184852 on Build GRCh38
Transcripts
NM_001105521.3 NM_001323086.1 NM_001323087.1 NM_001323088.1 NM_001323089.1 NM_001323090.1 XM_005252674.4 XM_005252676.4 XM_011539676.2 XM_011539682.2 XM_017016095.1 XM_017016096.1 XM_017016097.1 XM_017016098.1 XM_017016099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319347 c.1174C>G p.Q392E Chr.10: 132137076-132137076 on Build GRCh38
5370348 c.1151C>T p.S384F Chr.10: 132137053-132137053 on Build GRCh38
6689428 c.1215G>T p.V405V Chr.10: 132137117-132137117 on Build GRCh38
6689427 c.1145C>T p.P382L Chr.10: 132137047-132137047 on Build GRCh38
6689440 c.1197C>A p.F399L Chr.10: 132137099-132137099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747749806 Chr.10: 132137087-132137089 on Build GRCh38
rs371854189 Chr.10: 132137207-132137207 on Build GRCh38
rs201149274 Chr.10: 132137113-132137113 on Build GRCh38
rs750373773 Chr.10: 132137198-132137198 on Build GRCh38
rs751262268 Chr.10: 132137124-132137124 on Build GRCh38
rs999927477 Chr.10: 132137172-132137172 on Build GRCh38
rs773420827 Chr.10: 132137027-132137027 on Build GRCh38
rs995159690 Chr.10: 132137011-132137011 on Build GRCh38
rs967558386 Chr.10: 132137162-132137162 on Build GRCh38
rs547833056 Chr.10: 132137122-132137122 on Build GRCh38
rs781024453 Chr.10: 132137196-132137196 on Build GRCh38
rs201158135 Chr.10: 132137093-132137093 on Build GRCh38
rs1004311478 Chr.10: 132137077-132137077 on Build GRCh38
rs777099580 Chr.10: 132137175-132137176 on Build GRCh38
rs375263567 Chr.10: 132137222-132137222 on Build GRCh38
rs750605332 Chr.10: 132137080-132137080 on Build GRCh38
rs777791638 Chr.10: 132137228-132137228 on Build GRCh38
rs780921331 Chr.10: 132137136-132137136 on Build GRCh38
rs866725872 Chr.10: 132137132-132137133 on Build GRCh38
rs911671502 Chr.10: 132137171-132137171 on Build GRCh38
rs1016069275 Chr.10: 132137086-132137086 on Build GRCh38
rs772388290 Chr.10: 132137150-132137150 on Build GRCh38
rs1053273456 Chr.10: 132137159-132137159 on Build GRCh38
rs768911626 Chr.10: 132137139-132137139 on Build GRCh38
rs199828277 Chr.10: 132137041-132137041 on Build GRCh38
rs371025401 Chr.10: 132137097-132137097 on Build GRCh38
rs771430280 Chr.10: 132137142-132137145 on Build GRCh38
rs200516131 Chr.10: 132137180-132137180 on Build GRCh38
rs758979761 Chr.10: 132137066-132137066 on Build GRCh38
rs367725357 Chr.10: 132137178-132137178 on Build GRCh38
rs772618186 Chr.10: 132137021-132137021 on Build GRCh38
rs371845258 Chr.10: 132137173-132137173 on Build GRCh38
rs778695201 Chr.10: 132137023-132137024 on Build GRCh38
rs753403677 Chr.10: 132137064-132137064 on Build GRCh38
rs758013356 Chr.10: 132137117-132137117 on Build GRCh38
rs369946580 Chr.10: 132137199-132137199 on Build GRCh38
rs765240479 Chr.10: 132137016-132137016 on Build GRCh38
rs779116336 Chr.10: 132137144-132137144 on Build GRCh38
rs992865170 Chr.10: 132137118-132137118 on Build GRCh38
rs371392395 Chr.10: 132137040-132137040 on Build GRCh38
rs748423042 Chr.10: 132137146-132137146 on Build GRCh38
rs757229264 Chr.10: 132137134-132137134 on Build GRCh38
rs367921994 Chr.10: 132137156-132137156 on Build GRCh38
rs75996870 Chr.10: 132137138-132137138 on Build GRCh38
rs771259494 Chr.10: 132137235-132137235 on Build GRCh38
rs143120823 Chr.10: 132137174-132137174 on Build GRCh38
rs771474428 Chr.10: 132137157-132137157 on Build GRCh38
rs774777576 Chr.10: 132137012-132137012 on Build GRCh38
rs372686219 Chr.10: 132137179-132137179 on Build GRCh38
rs539382663 Chr.10: 132137182-132137182 on Build GRCh38
rs960090419 Chr.10: 132137114-132137114 on Build GRCh38
rs866030750 Chr.10: 132137017-132137017 on Build GRCh38
rs533727645 Chr.10: 132137160-132137160 on Build GRCh38
rs747252700 Chr.10: 132137229-132137229 on Build GRCh38
rs1034845722 Chr.10: 132137203-132137203 on Build GRCh38
rs34903225 Chr.10: 132137199-132137199 on Build GRCh38

View Full Product Details