Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101961665-101961938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP126
Entrez Gene ID
57562
Gene Name
centrosomal protein 126
Gene Aliases
KIAA1377
UniGene
Hs.156352
Gene Chromosome Location
Chr.11: 101915015-102001065 on Build GRCh38
Transcripts
NM_020802.3 XM_005271627.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417598 c.842T>C p.I281T Chr.11: 101961877-101961877 on Build GRCh38
4417522 c.824C>A p.S275Y Chr.11: 101961859-101961859 on Build GRCh38
5969967 c.723T>C p.V241V Chr.11: 101961758-101961758 on Build GRCh38
1127955 c.823T>C p.S275P Chr.11: 101961858-101961858 on Build GRCh38
2107950 c.869C>T p.S290L Chr.11: 101961904-101961904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903740664 Chr.11: 101961806-101961806 on Build GRCh38
rs757650654 Chr.11: 101961897-101961897 on Build GRCh38
rs768130043 Chr.11: 101961857-101961857 on Build GRCh38
rs564898739 Chr.11: 101961728-101961728 on Build GRCh38
rs145509374 Chr.11: 101961890-101961890 on Build GRCh38
rs11225089 Chr.11: 101961859-101961859 on Build GRCh38
rs768472802 Chr.11: 101961730-101961730 on Build GRCh38
rs751801624 Chr.11: 101961810-101961810 on Build GRCh38
rs774236498 Chr.11: 101961871-101961871 on Build GRCh38
rs755212069 Chr.11: 101961815-101961815 on Build GRCh38
rs777651964 Chr.11: 101961727-101961728 on Build GRCh38
rs7926728 Chr.11: 101961747-101961747 on Build GRCh38
rs760281171 Chr.11: 101961722-101961722 on Build GRCh38
rs374315228 Chr.11: 101961774-101961774 on Build GRCh38
rs749839838 Chr.11: 101961836-101961836 on Build GRCh38
rs866252257 Chr.11: 101961812-101961812 on Build GRCh38
rs778200856 Chr.11: 101961834-101961834 on Build GRCh38
rs955585588 Chr.11: 101961853-101961853 on Build GRCh38
rs779481661 Chr.11: 101961898-101961898 on Build GRCh38
rs766435725 Chr.11: 101961783-101961783 on Build GRCh38
rs369919672 Chr.11: 101961912-101961912 on Build GRCh38
rs375546304 Chr.11: 101961868-101961868 on Build GRCh38
rs769761349 Chr.11: 101961860-101961860 on Build GRCh38
rs905613821 Chr.11: 101961786-101961786 on Build GRCh38
rs867558304 Chr.11: 101961758-101961758 on Build GRCh38
rs11225090 Chr.11: 101961877-101961877 on Build GRCh38
rs147605435 Chr.11: 101961849-101961849 on Build GRCh38
rs779492169 Chr.11: 101961842-101961842 on Build GRCh38
rs541103749 Chr.11: 101961728-101961729 on Build GRCh38
rs752936061 Chr.11: 101961881-101961881 on Build GRCh38
rs376836980 Chr.11: 101961714-101961714 on Build GRCh38
rs781489478 Chr.11: 101961825-101961825 on Build GRCh38
rs773254227 Chr.11: 101961861-101961861 on Build GRCh38
rs747982374 Chr.11: 101961706-101961707 on Build GRCh38
rs752991138 Chr.11: 101961828-101961828 on Build GRCh38
rs756499304 Chr.11: 101961833-101961833 on Build GRCh38
rs145875462 Chr.11: 101961767-101961767 on Build GRCh38
rs776456092 Chr.11: 101961858-101961858 on Build GRCh38
rs774574322 Chr.11: 101961896-101961896 on Build GRCh38
rs776515192 Chr.11: 101961736-101961736 on Build GRCh38
rs145307984 Chr.11: 101961760-101961760 on Build GRCh38
rs757926755 Chr.11: 101961837-101961837 on Build GRCh38
rs542965138 Chr.11: 101961867-101961867 on Build GRCh38
rs1001270427 Chr.11: 101961798-101961798 on Build GRCh38
rs1052085449 Chr.11: 101961856-101961856 on Build GRCh38
rs746464213 Chr.11: 101961903-101961903 on Build GRCh38
rs771282935 Chr.11: 101961887-101961887 on Build GRCh38
rs561312521 Chr.11: 101961707-101961707 on Build GRCh38
rs746980072 Chr.11: 101961706-101961706 on Build GRCh38
rs1043658866 Chr.11: 101961804-101961804 on Build GRCh38

View Full Product Details