Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81659284-81659553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPA1
Entrez Gene ID
3612
Gene Name
inositol monophosphatase 1
Gene Aliases
IMP, IMPA
UniGene
Hs.656694
Gene Chromosome Location
Chr.8: 81656916-81686354 on Build GRCh38
Transcripts
NM_001144878.1 NM_001144879.1 NM_005536.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3651325 c.782G>A p.R261K Chr.8: 81659403-81659403 on Build GRCh38
2876993 c.743G>A p.R248Q Chr.8: 81659442-81659442 on Build GRCh38
6181933 c.970G>C p.E324Q Chr.8: 81659392-81659392 on Build GRCh38
2876991 c.798T>C p.I266I Chr.8: 81659387-81659387 on Build GRCh38
3651324 c.959G>A p.R320K Chr.8: 81659403-81659403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778160191 Chr.8: 81659443-81659443 on Build GRCh38
rs759673501 Chr.8: 81659494-81659494 on Build GRCh38
rs764082454 Chr.8: 81659369-81659369 on Build GRCh38
rs372313423 Chr.8: 81659363-81659363 on Build GRCh38
rs745483441 Chr.8: 81659327-81659327 on Build GRCh38
rs921267572 Chr.8: 81659521-81659521 on Build GRCh38
rs963555944 Chr.8: 81659496-81659496 on Build GRCh38
rs773742914 Chr.8: 81659355-81659355 on Build GRCh38
rs770239191 Chr.8: 81659352-81659352 on Build GRCh38
rs146624224 Chr.8: 81659430-81659430 on Build GRCh38
rs775811695 Chr.8: 81659514-81659514 on Build GRCh38
rs768524208 Chr.8: 81659355-81659356 on Build GRCh38
rs778270892 Chr.8: 81659401-81659401 on Build GRCh38
rs767783535 Chr.8: 81659362-81659362 on Build GRCh38
rs776798105 Chr.8: 81659330-81659330 on Build GRCh38
rs766424862 Chr.8: 81659357-81659357 on Build GRCh38
rs774228187 Chr.8: 81659463-81659463 on Build GRCh38
rs374985214 Chr.8: 81659398-81659398 on Build GRCh38
rs749781827 Chr.8: 81659446-81659446 on Build GRCh38
rs771496191 Chr.8: 81659447-81659447 on Build GRCh38
rs753792061 Chr.8: 81659386-81659386 on Build GRCh38
rs10105165 Chr.8: 81659344-81659344 on Build GRCh38
rs771811787 Chr.8: 81659328-81659328 on Build GRCh38
rs772136390 Chr.8: 81659502-81659502 on Build GRCh38
rs779780582 Chr.8: 81659437-81659437 on Build GRCh38
rs144509656 Chr.8: 81659359-81659359 on Build GRCh38
rs928457327 Chr.8: 81659332-81659332 on Build GRCh38
rs770116448 Chr.8: 81659442-81659442 on Build GRCh38
rs755994131 Chr.8: 81659367-81659367 on Build GRCh38
rs368377674 Chr.8: 81659331-81659331 on Build GRCh38
rs745447728 Chr.8: 81659422-81659422 on Build GRCh38
rs139924114 Chr.8: 81659438-81659438 on Build GRCh38
rs1004935117 Chr.8: 81659376-81659376 on Build GRCh38

View Full Product Details