Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 29919886-29920145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL1
Entrez Gene ID
11141
Gene Name
interleukin 1 receptor accessory protein like 1
Gene Aliases
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2
UniGene
Hs.658912
Gene Chromosome Location
Chr.X: 28587564-29956350 on Build GRCh38
Transcripts
NM_014271.3 XM_017029240.1 XM_017029241.1

SNP Detail

SNP ID Chromosome Location
rs183925588 Chr.X: 29920084-29920084 on Build GRCh38
rs199971765 Chr.X: 29920063-29920063 on Build GRCh38
rs779866333 Chr.X: 29920062-29920062 on Build GRCh38
rs1009288428 Chr.X: 29919929-29919929 on Build GRCh38
rs371275154 Chr.X: 29920072-29920072 on Build GRCh38
rs372256169 Chr.X: 29920118-29920118 on Build GRCh38
rs749380359 Chr.X: 29920070-29920070 on Build GRCh38
rs369557817 Chr.X: 29920075-29920075 on Build GRCh38
rs756426427 Chr.X: 29920065-29920065 on Build GRCh38
rs770839799 Chr.X: 29920074-29920074 on Build GRCh38
rs747300297 Chr.X: 29920092-29920092 on Build GRCh38
rs778100377 Chr.X: 29920067-29920067 on Build GRCh38
rs758196137 Chr.X: 29919957-29919957 on Build GRCh38
rs749383705 Chr.X: 29920117-29920117 on Build GRCh38
rs772374800 Chr.X: 29920079-29920079 on Build GRCh38
rs200419221 Chr.X: 29920076-29920076 on Build GRCh38
rs995057356 Chr.X: 29920061-29920061 on Build GRCh38

View Full Product Details