Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92223151-92223424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
UniGene
Hs.445403
Gene Chromosome Location
Chr.9: 92210207-92293756 on Build GRCh38
Transcripts
NM_002161.5 NM_013417.3 NR_073446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
289531 c.3544A>C p.K1182Q Chr.9: 92223355-92223355 on Build GRCh38
7186931 c.3520A>G p.I1174V Chr.9: 92223379-92223379 on Build GRCh38
6298852 c.3553+8C>A p.? Chr.9: 92223338-92223338 on Build GRCh38
1463559 c.3524A>G p.N1175S Chr.9: 92223375-92223375 on Build GRCh38
3764060 c.3544A>G p.K1182E Chr.9: 92223355-92223355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370274531 Chr.9: 92223350-92223350 on Build GRCh38
rs375173284 Chr.9: 92223312-92223312 on Build GRCh38
rs749373708 Chr.9: 92223348-92223348 on Build GRCh38
rs746959650 Chr.9: 92223358-92223358 on Build GRCh38
rs200405574 Chr.9: 92223313-92223313 on Build GRCh38
rs779253484 Chr.9: 92223359-92223359 on Build GRCh38
rs756131376 Chr.9: 92223233-92223233 on Build GRCh38
rs757227229 Chr.9: 92223223-92223223 on Build GRCh38
rs898518787 Chr.9: 92223194-92223194 on Build GRCh38
rs747772656 Chr.9: 92223396-92223396 on Build GRCh38
rs374633832 Chr.9: 92223400-92223400 on Build GRCh38
rs72750411 Chr.9: 92223246-92223246 on Build GRCh38
rs886037873 Chr.9: 92223378-92223378 on Build GRCh38
rs866942945 Chr.9: 92223259-92223259 on Build GRCh38
rs185921740 Chr.9: 92223297-92223297 on Build GRCh38
rs752901332 Chr.9: 92223332-92223332 on Build GRCh38
rs879413348 Chr.9: 92223213-92223213 on Build GRCh38
rs1040238198 Chr.9: 92223244-92223244 on Build GRCh38
rs966664738 Chr.9: 92223285-92223285 on Build GRCh38
rs62565125 Chr.9: 92223254-92223254 on Build GRCh38
rs143955291 Chr.9: 92223191-92223191 on Build GRCh38
rs777875841 Chr.9: 92223347-92223347 on Build GRCh38
rs891233364 Chr.9: 92223230-92223231 on Build GRCh38
rs998151029 Chr.9: 92223260-92223260 on Build GRCh38
rs146425870 Chr.9: 92223204-92223204 on Build GRCh38
rs750934877 Chr.9: 92223180-92223180 on Build GRCh38
rs370962208 Chr.9: 92223279-92223279 on Build GRCh38
rs755268664 Chr.9: 92223315-92223315 on Build GRCh38
rs556155 Chr.9: 92223355-92223355 on Build GRCh38
rs749172818 Chr.9: 92223174-92223174 on Build GRCh38
rs754623291 Chr.9: 92223253-92223253 on Build GRCh38
rs1032426613 Chr.9: 92223343-92223343 on Build GRCh38
rs556104028 Chr.9: 92223235-92223235 on Build GRCh38
rs768561823 Chr.9: 92223373-92223373 on Build GRCh38
rs767881151 Chr.9: 92223316-92223316 on Build GRCh38
rs772640056 Chr.9: 92223300-92223300 on Build GRCh38
rs929741713 Chr.9: 92223203-92223203 on Build GRCh38
rs999614574 Chr.9: 92223286-92223286 on Build GRCh38
rs560795216 Chr.9: 92223381-92223381 on Build GRCh38
rs780132311 Chr.9: 92223245-92223245 on Build GRCh38
rs756392976 Chr.9: 92223337-92223337 on Build GRCh38
rs144431665 Chr.9: 92223379-92223379 on Build GRCh38

View Full Product Details