Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139166586-139166859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828586 c.725G>A p.R242Q Chr.6: 139166737-139166737 on Build GRCh38
3829172 c.621C>T p.S207S Chr.6: 139166633-139166633 on Build GRCh38
3777155 c.643C>T p.P215S Chr.6: 139166655-139166655 on Build GRCh38
4793006 c.623A>T p.E208V Chr.6: 139166635-139166635 on Build GRCh38
5923725 c.641C>T p.P214L Chr.6: 139166653-139166653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748644903 Chr.6: 139166796-139166796 on Build GRCh38
rs754119417 Chr.6: 139166633-139166633 on Build GRCh38
rs772720494 Chr.6: 139166624-139166624 on Build GRCh38
rs747861281 Chr.6: 139166686-139166686 on Build GRCh38
rs200446030 Chr.6: 139166799-139166799 on Build GRCh38
rs776074028 Chr.6: 139166627-139166627 on Build GRCh38
rs755906012 Chr.6: 139166756-139166756 on Build GRCh38
rs202156473 Chr.6: 139166687-139166687 on Build GRCh38
rs762507172 Chr.6: 139166610-139166610 on Build GRCh38
rs777075681 Chr.6: 139166716-139166716 on Build GRCh38
rs765446222 Chr.6: 139166727-139166727 on Build GRCh38
rs770198210 Chr.6: 139166798-139166798 on Build GRCh38
rs573560322 Chr.6: 139166800-139166800 on Build GRCh38
rs764579603 Chr.6: 139166632-139166632 on Build GRCh38
rs1009332556 Chr.6: 139166823-139166823 on Build GRCh38
rs913811849 Chr.6: 139166737-139166737 on Build GRCh38
rs762125294 Chr.6: 139166638-139166638 on Build GRCh38
rs766550253 Chr.6: 139166801-139166801 on Build GRCh38
rs868585045 Chr.6: 139166775-139166775 on Build GRCh38
rs778514652 Chr.6: 139166781-139166781 on Build GRCh38
rs752298876 Chr.6: 139166677-139166677 on Build GRCh38
rs751564590 Chr.6: 139166736-139166736 on Build GRCh38
rs1002457088 Chr.6: 139166805-139166805 on Build GRCh38
rs773487294 Chr.6: 139166729-139166729 on Build GRCh38
rs758402074 Chr.6: 139166665-139166665 on Build GRCh38
rs748813213 Chr.6: 139166695-139166695 on Build GRCh38
rs774515194 Chr.6: 139166804-139166804 on Build GRCh38
rs557021529 Chr.6: 139166642-139166642 on Build GRCh38
rs777706849 Chr.6: 139166761-139166761 on Build GRCh38
rs1032927797 Chr.6: 139166809-139166809 on Build GRCh38
rs3173082 Chr.6: 139166699-139166699 on Build GRCh38
rs756949496 Chr.6: 139166770-139166770 on Build GRCh38
rs754847006 Chr.6: 139166672-139166672 on Build GRCh38
rs769303187 Chr.6: 139166609-139166609 on Build GRCh38
rs776935770 Chr.6: 139166793-139166793 on Build GRCh38
rs548239720 Chr.6: 139166836-139166836 on Build GRCh38
rs747594392 Chr.6: 139166616-139166618 on Build GRCh38
rs145902743 Chr.6: 139166649-139166649 on Build GRCh38
rs766492805 Chr.6: 139166733-139166733 on Build GRCh38
rs893941207 Chr.6: 139166819-139166819 on Build GRCh38
rs376482952 Chr.6: 139166668-139166668 on Build GRCh38
rs762026133 Chr.6: 139166723-139166723 on Build GRCh38
rs921128357 Chr.6: 139166660-139166660 on Build GRCh38
rs370979954 Chr.6: 139166669-139166669 on Build GRCh38
rs200173389 Chr.6: 139166675-139166675 on Build GRCh38
rs763145853 Chr.6: 139166731-139166731 on Build GRCh38
rs529694661 Chr.6: 139166783-139166783 on Build GRCh38
rs775887767 Chr.6: 139166666-139166666 on Build GRCh38
rs761084053 Chr.6: 139166630-139166630 on Build GRCh38
rs770633229 Chr.6: 139166696-139166696 on Build GRCh38
rs767312064 Chr.6: 139166752-139166752 on Build GRCh38
rs528690442 Chr.6: 139166692-139166692 on Build GRCh38
rs931389617 Chr.6: 139166766-139166766 on Build GRCh38
rs146626982 Chr.6: 139166751-139166751 on Build GRCh38
rs368450813 Chr.6: 139166792-139166792 on Build GRCh38
rs769002874 Chr.6: 139166701-139166701 on Build GRCh38
rs749007568 Chr.6: 139166765-139166765 on Build GRCh38
rs773723392 Chr.6: 139166697-139166697 on Build GRCh38
rs767495347 Chr.6: 139166838-139166838 on Build GRCh38

View Full Product Details