Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138182825-138183078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF20A
Entrez Gene ID
10112
Gene Name
kinesin family member 20A
Gene Aliases
MKLP2, RAB6KIFL
UniGene
Hs.718626
Gene Chromosome Location
Chr.5: 138178728-138187715 on Build GRCh38
Transcripts
NM_005733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428145 c.803G>A p.S268N Chr.5: 138182961-138182961 on Build GRCh38
3826833 c.784G>C p.A262P Chr.5: 138182942-138182942 on Build GRCh38
735572 c.800T>C p.I267T Chr.5: 138182958-138182958 on Build GRCh38
141120 c.728G>A p.R243K Chr.5: 138182886-138182886 on Build GRCh38
5154166 c.822C>T p.S274S Chr.5: 138182980-138182980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748238660 Chr.5: 138182980-138182980 on Build GRCh38
rs773275190 Chr.5: 138182906-138182906 on Build GRCh38
rs766215564 Chr.5: 138182908-138182908 on Build GRCh38
rs144853219 Chr.5: 138182899-138182899 on Build GRCh38
rs571617605 Chr.5: 138182874-138182874 on Build GRCh38
rs781714038 Chr.5: 138182930-138182930 on Build GRCh38
rs201018610 Chr.5: 138183009-138183009 on Build GRCh38
rs767362355 Chr.5: 138183030-138183033 on Build GRCh38
rs532811153 Chr.5: 138182896-138182896 on Build GRCh38
rs1013611042 Chr.5: 138183021-138183021 on Build GRCh38
rs750863843 Chr.5: 138182937-138182937 on Build GRCh38
rs764911233 Chr.5: 138182922-138182922 on Build GRCh38
rs781648104 Chr.5: 138182854-138182854 on Build GRCh38
rs759068632 Chr.5: 138182917-138182917 on Build GRCh38
rs768806257 Chr.5: 138182884-138182884 on Build GRCh38
rs112621273 Chr.5: 138183001-138183001 on Build GRCh38
rs779178029 Chr.5: 138182975-138182975 on Build GRCh38
rs752197079 Chr.5: 138182923-138182923 on Build GRCh38
rs751217694 Chr.5: 138183015-138183016 on Build GRCh38
rs746290122 Chr.5: 138182859-138182859 on Build GRCh38
rs763809159 Chr.5: 138182859-138182861 on Build GRCh38
rs768804633 Chr.5: 138182971-138182971 on Build GRCh38
rs764855719 Chr.5: 138183008-138183008 on Build GRCh38
rs775696409 Chr.5: 138182865-138182865 on Build GRCh38
rs757744529 Chr.5: 138182853-138182853 on Build GRCh38
rs761581021 Chr.5: 138183024-138183025 on Build GRCh38
rs947005155 Chr.5: 138183054-138183054 on Build GRCh38
rs138686481 Chr.5: 138183000-138183000 on Build GRCh38
rs760546603 Chr.5: 138182907-138182907 on Build GRCh38
rs374368372 Chr.5: 138183041-138183041 on Build GRCh38
rs941036797 Chr.5: 138182934-138182934 on Build GRCh38
rs200525885 Chr.5: 138182900-138182900 on Build GRCh38
rs756486916 Chr.5: 138182947-138182947 on Build GRCh38
rs377038296 Chr.5: 138182986-138182986 on Build GRCh38
rs760638384 Chr.5: 138182991-138182991 on Build GRCh38
rs780350277 Chr.5: 138182959-138182959 on Build GRCh38
rs753548639 Chr.5: 138182910-138182910 on Build GRCh38
rs73261939 Chr.5: 138182901-138182901 on Build GRCh38
rs757890608 Chr.5: 138182929-138182929 on Build GRCh38
rs373041750 Chr.5: 138182864-138182864 on Build GRCh38
rs759352647 Chr.5: 138183007-138183007 on Build GRCh38
rs770878372 Chr.5: 138182999-138182999 on Build GRCh38

View Full Product Details