Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101962035-101962262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP126
Entrez Gene ID
57562
Gene Name
centrosomal protein 126
Gene Aliases
KIAA1377
UniGene
Hs.156352
Gene Chromosome Location
Chr.11: 101915015-102001065 on Build GRCh38
Transcripts
NM_020802.3 XM_005271627.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657695 c.1176G>A p.M392I Chr.11: 101962211-101962211 on Build GRCh38
6067029 c.1148G>C p.C383S Chr.11: 101962183-101962183 on Build GRCh38
5922895 c.1001C>T p.S334F Chr.11: 101962036-101962036 on Build GRCh38
5909121 c.1049C>T p.P350L Chr.11: 101962084-101962084 on Build GRCh38
4457672 c.1054C>T p.P352S Chr.11: 101962089-101962089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767006537 Chr.11: 101962173-101962173 on Build GRCh38
rs560892333 Chr.11: 101962127-101962127 on Build GRCh38
rs763613769 Chr.11: 101962160-101962160 on Build GRCh38
rs369177809 Chr.11: 101962230-101962230 on Build GRCh38
rs777516747 Chr.11: 101962204-101962204 on Build GRCh38
rs752186613 Chr.11: 101962191-101962191 on Build GRCh38
rs777571650 Chr.11: 101962087-101962087 on Build GRCh38
rs775494260 Chr.11: 101962141-101962141 on Build GRCh38
rs369939612 Chr.11: 101962150-101962150 on Build GRCh38
rs373160620 Chr.11: 101962209-101962209 on Build GRCh38
rs377438714 Chr.11: 101962223-101962223 on Build GRCh38
rs61745759 Chr.11: 101962149-101962149 on Build GRCh38
rs780916857 Chr.11: 101962078-101962078 on Build GRCh38
rs772129915 Chr.11: 101962139-101962139 on Build GRCh38
rs145904743 Chr.11: 101962094-101962094 on Build GRCh38
rs756968310 Chr.11: 101962210-101962210 on Build GRCh38
rs893326654 Chr.11: 101962088-101962088 on Build GRCh38
rs778690053 Chr.11: 101962111-101962111 on Build GRCh38
rs778831481 Chr.11: 101962214-101962214 on Build GRCh38
rs745854138 Chr.11: 101962118-101962118 on Build GRCh38
rs780000406 Chr.11: 101962228-101962228 on Build GRCh38
rs750891461 Chr.11: 101962154-101962154 on Build GRCh38
rs1052558937 Chr.11: 101962183-101962183 on Build GRCh38
rs755768726 Chr.11: 101962083-101962083 on Build GRCh38
rs770779618 Chr.11: 101962104-101962104 on Build GRCh38
rs752384562 Chr.11: 101962081-101962081 on Build GRCh38
rs890777327 Chr.11: 101962211-101962211 on Build GRCh38
rs935205479 Chr.11: 101962172-101962172 on Build GRCh38
rs768830068 Chr.11: 101962144-101962144 on Build GRCh38
rs765676576 Chr.11: 101962151-101962151 on Build GRCh38
rs745775888 Chr.11: 101962215-101962215 on Build GRCh38
rs1020368425 Chr.11: 101962129-101962129 on Build GRCh38

View Full Product Details