Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176130840-176131065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD8
Entrez Gene ID
3234
Gene Name
homeobox D8
Gene Aliases
HOX4, HOX4E, HOX5.4
UniGene
Hs.301963
Gene Chromosome Location
Chr.2: 176129694-176132695 on Build GRCh38
Transcripts
NM_001199746.1 NM_001199747.1 NM_019558.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5973418 c.512A>G p.E171G Chr.2: 176130878-176130878 on Build GRCh38
7215940 c.513G>A p.E171E Chr.2: 176130879-176130879 on Build GRCh38
3743348 c.477T>C p.Y159Y Chr.2: 176130843-176130843 on Build GRCh38
5699608 c.535A>G p.S179G Chr.2: 176130901-176130901 on Build GRCh38
441537 c.550C>G p.Q184E Chr.2: 176130916-176130916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777009071 Chr.2: 176130955-176130955 on Build GRCh38
rs142220242 Chr.2: 176130926-176130926 on Build GRCh38
rs767574519 Chr.2: 176130992-176130992 on Build GRCh38
rs754952797 Chr.2: 176130905-176130905 on Build GRCh38
rs754471508 Chr.2: 176130980-176130980 on Build GRCh38
rs769151115 Chr.2: 176130954-176130954 on Build GRCh38
rs1036380783 Chr.2: 176130912-176130912 on Build GRCh38
rs775411258 Chr.2: 176130935-176130935 on Build GRCh38
rs757955465 Chr.2: 176130886-176130886 on Build GRCh38
rs371171284 Chr.2: 176130933-176130933 on Build GRCh38
rs912201059 Chr.2: 176131029-176131029 on Build GRCh38
rs939782863 Chr.2: 176131032-176131032 on Build GRCh38
rs751109255 Chr.2: 176130978-176130978 on Build GRCh38
rs1036701798 Chr.2: 176131034-176131034 on Build GRCh38
rs771423872 Chr.2: 176130911-176130913 on Build GRCh38
rs754192277 Chr.2: 176130866-176130866 on Build GRCh38
rs1024106161 Chr.2: 176130990-176130990 on Build GRCh38
rs777057976 Chr.2: 176130960-176130960 on Build GRCh38
rs577550151 Chr.2: 176131002-176131002 on Build GRCh38
rs746039102 Chr.2: 176130924-176130924 on Build GRCh38
rs748445815 Chr.2: 176130999-176130999 on Build GRCh38
rs138972805 Chr.2: 176130906-176130906 on Build GRCh38
rs762340147 Chr.2: 176130954-176130955 on Build GRCh38
rs749992151 Chr.2: 176130884-176130884 on Build GRCh38
rs148123943 Chr.2: 176130891-176130891 on Build GRCh38
rs765476275 Chr.2: 176130966-176130966 on Build GRCh38
rs898039591 Chr.2: 176131037-176131037 on Build GRCh38
rs762181268 Chr.2: 176130965-176130965 on Build GRCh38
rs538373838 Chr.2: 176131009-176131009 on Build GRCh38
rs779395863 Chr.2: 176130888-176130888 on Build GRCh38
rs1014105480 Chr.2: 176130973-176130973 on Build GRCh38
rs3765049 Chr.2: 176130879-176130879 on Build GRCh38
rs552971699 Chr.2: 176130975-176130975 on Build GRCh38
rs747959114 Chr.2: 176130910-176130910 on Build GRCh38
rs761522615 Chr.2: 176130878-176130878 on Build GRCh38
rs199981512 Chr.2: 176130967-176130967 on Build GRCh38
rs556612303 Chr.2: 176131036-176131036 on Build GRCh38
rs752634170 Chr.2: 176130995-176130995 on Build GRCh38
rs373727289 Chr.2: 176130949-176130949 on Build GRCh38
rs765541743 Chr.2: 176130972-176130973 on Build GRCh38
rs555382258 Chr.2: 176130918-176130918 on Build GRCh38

View Full Product Details