Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133074030-133074255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA4
Entrez Gene ID
3308
Gene Name
heat shock protein family A (Hsp70) member 4
Gene Aliases
APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY
UniGene
Hs.90093
Gene Chromosome Location
Chr.5: 133051970-133105017 on Build GRCh38
Transcripts
NM_002154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060807 c.646A>G p.N216D Chr.5: 133074109-133074109 on Build GRCh38
1060808 c.655A>T p.K219* Chr.5: 133074118-133074118 on Build GRCh38
5402119 c.598T>G p.F200V Chr.5: 133074061-133074061 on Build GRCh38
1060806 c.589A>T p.N197Y Chr.5: 133074052-133074052 on Build GRCh38
6168653 c.632C>T p.S211F Chr.5: 133074095-133074095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115939709 Chr.5: 133074057-133074057 on Build GRCh38
rs762762736 Chr.5: 133074124-133074124 on Build GRCh38
rs540903929 Chr.5: 133074137-133074138 on Build GRCh38
rs749967827 Chr.5: 133074147-133074147 on Build GRCh38
rs925103222 Chr.5: 133074209-133074209 on Build GRCh38
rs989695812 Chr.5: 133074182-133074182 on Build GRCh38
rs543333439 Chr.5: 133074227-133074227 on Build GRCh38
rs958355620 Chr.5: 133074176-133074176 on Build GRCh38
rs747218789 Chr.5: 133074170-133074170 on Build GRCh38
rs374749021 Chr.5: 133074150-133074150 on Build GRCh38
rs755480705 Chr.5: 133074148-133074148 on Build GRCh38
rs751220191 Chr.5: 133074135-133074135 on Build GRCh38
rs779512301 Chr.5: 133074155-133074156 on Build GRCh38
rs112014979 Chr.5: 133074212-133074212 on Build GRCh38
rs186579297 Chr.5: 133074165-133074165 on Build GRCh38
rs371614765 Chr.5: 133074096-133074096 on Build GRCh38
rs761171742 Chr.5: 133074137-133074137 on Build GRCh38
rs763699104 Chr.5: 133074133-133074133 on Build GRCh38
rs62375235 Chr.5: 133074175-133074175 on Build GRCh38
rs913571335 Chr.5: 133074215-133074215 on Build GRCh38
rs759508272 Chr.5: 133074070-133074070 on Build GRCh38
rs767041927 Chr.5: 133074145-133074145 on Build GRCh38
rs35067922 Chr.5: 133074096-133074097 on Build GRCh38
rs368824221 Chr.5: 133074171-133074171 on Build GRCh38
rs755471375 Chr.5: 133074138-133074138 on Build GRCh38
rs775246521 Chr.5: 133074082-133074082 on Build GRCh38
rs1006411916 Chr.5: 133074152-133074152 on Build GRCh38
rs976366134 Chr.5: 133074225-133074225 on Build GRCh38
rs753071388 Chr.5: 133074153-133074153 on Build GRCh38
rs897837514 Chr.5: 133074105-133074105 on Build GRCh38
rs758836948 Chr.5: 133074164-133074164 on Build GRCh38
rs1052347768 Chr.5: 133074214-133074214 on Build GRCh38
rs952338056 Chr.5: 133074146-133074146 on Build GRCh38
rs993468209 Chr.5: 133074138-133074138 on Build GRCh38
rs140131673 Chr.5: 133074184-133074184 on Build GRCh38
rs769655551 Chr.5: 133074071-133074071 on Build GRCh38

View Full Product Details