Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108191450-108191723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT57
Entrez Gene ID
55081
Gene Name
intraflagellar transport 57
Gene Aliases
ESRRBL1, HIPPI, MHS4R2
UniGene
Hs.412196
Gene Chromosome Location
Chr.3: 108160812-108222570 on Build GRCh38
Transcripts
NM_018010.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036014 c.663C>T p.N221N Chr.3: 108191635-108191635 on Build GRCh38
3737539 c.760A>T p.I254F Chr.3: 108191538-108191538 on Build GRCh38
5863158 c.743C>T p.P248L Chr.3: 108191555-108191555 on Build GRCh38
1036013 c.758C>T p.T253M Chr.3: 108191540-108191540 on Build GRCh38
5059182 c.728T>G p.V243G Chr.3: 108191570-108191570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375684447 Chr.3: 108191616-108191616 on Build GRCh38
rs752845506 Chr.3: 108191555-108191555 on Build GRCh38
rs767055929 Chr.3: 108191482-108191482 on Build GRCh38
rs778811894 Chr.3: 108191497-108191497 on Build GRCh38
rs749923537 Chr.3: 108191552-108191552 on Build GRCh38
rs758385329 Chr.3: 108191571-108191571 on Build GRCh38
rs182941979 Chr.3: 108191635-108191635 on Build GRCh38
rs764078055 Chr.3: 108191668-108191668 on Build GRCh38
rs143872284 Chr.3: 108191659-108191659 on Build GRCh38
rs752295540 Chr.3: 108191481-108191481 on Build GRCh38
rs544087097 Chr.3: 108191498-108191498 on Build GRCh38
rs142978591 Chr.3: 108191586-108191586 on Build GRCh38
rs780363078 Chr.3: 108191596-108191596 on Build GRCh38
rs370528818 Chr.3: 108191550-108191550 on Build GRCh38
rs755224848 Chr.3: 108191678-108191678 on Build GRCh38
rs796918388 Chr.3: 108191481-108191482 on Build GRCh38
rs775640428 Chr.3: 108191520-108191520 on Build GRCh38
rs753603293 Chr.3: 108191494-108191494 on Build GRCh38
rs932265377 Chr.3: 108191662-108191662 on Build GRCh38
rs575087163 Chr.3: 108191487-108191487 on Build GRCh38
rs753425077 Chr.3: 108191641-108191641 on Build GRCh38
rs774931041 Chr.3: 108191540-108191540 on Build GRCh38
rs371129548 Chr.3: 108191565-108191565 on Build GRCh38
rs60290956 Chr.3: 108191494-108191495 on Build GRCh38
rs759926220 Chr.3: 108191480-108191481 on Build GRCh38
rs752592683 Chr.3: 108191569-108191569 on Build GRCh38
rs913317756 Chr.3: 108191597-108191597 on Build GRCh38
rs1004533497 Chr.3: 108191514-108191514 on Build GRCh38
rs375744646 Chr.3: 108191554-108191554 on Build GRCh38
rs769154579 Chr.3: 108191533-108191533 on Build GRCh38
rs779618783 Chr.3: 108191492-108191492 on Build GRCh38
rs1029207576 Chr.3: 108191563-108191563 on Build GRCh38
rs757073831 Chr.3: 108191481-108191483 on Build GRCh38
rs746028787 Chr.3: 108191508-108191508 on Build GRCh38
rs749468576 Chr.3: 108191676-108191676 on Build GRCh38
rs532732144 Chr.3: 108191481-108191482 on Build GRCh38
rs147450354 Chr.3: 108191601-108191601 on Build GRCh38
rs753551661 Chr.3: 108191557-108191557 on Build GRCh38
rs770932039 Chr.3: 108191505-108191505 on Build GRCh38
rs769821703 Chr.3: 108191590-108191590 on Build GRCh38
rs377596960 Chr.3: 108191524-108191524 on Build GRCh38
rs199827942 Chr.3: 108191665-108191665 on Build GRCh38
rs754838491 Chr.3: 108191496-108191496 on Build GRCh38
rs202070886 Chr.3: 108191506-108191506 on Build GRCh38
rs769099346 Chr.3: 108191614-108191614 on Build GRCh38
rs754712026 Chr.3: 108191564-108191564 on Build GRCh38
rs900493736 Chr.3: 108191661-108191661 on Build GRCh38
rs751464539 Chr.3: 108191669-108191669 on Build GRCh38
rs139075839 Chr.3: 108191579-108191579 on Build GRCh38
rs372553451 Chr.3: 108191513-108191513 on Build GRCh38
rs201461104 Chr.3: 108191675-108191675 on Build GRCh38
rs371760728 Chr.3: 108191491-108191491 on Build GRCh38
rs867145435 Chr.3: 108191644-108191644 on Build GRCh38
rs772596396 Chr.3: 108191628-108191628 on Build GRCh38
rs764964944 Chr.3: 108191652-108191652 on Build GRCh38
rs753930289 Chr.3: 108191634-108191634 on Build GRCh38
rs762312928 Chr.3: 108191626-108191626 on Build GRCh38
rs759218939 Chr.3: 108191650-108191650 on Build GRCh38

View Full Product Details