Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151437187-151437440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF10
Entrez Gene ID
285313
Gene Name
immunoglobulin superfamily member 10
Gene Aliases
CMF608
UniGene
Hs.58561
Gene Chromosome Location
Chr.3: 151435051-151619926 on Build GRCh38
Transcripts
NM_001178145.1 NM_001178146.1 NM_178822.4 XM_011512708.2 XM_011512709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5834566 c.7127delG p.G2376fs*14 Chr.3: 151437434-151437434 on Build GRCh38
1210395 c.7132C>T p.R2378* Chr.3: 151437429-151437429 on Build GRCh38
4999705 c.7142A>G p.N2381S Chr.3: 151437419-151437419 on Build GRCh38
7216363 c.7311T>C p.Y2437Y Chr.3: 151437250-151437250 on Build GRCh38
4506826 c.7303C>T p.L2435F Chr.3: 151437258-151437258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044188598 Chr.3: 151437351-151437351 on Build GRCh38
rs144086705 Chr.3: 151437288-151437288 on Build GRCh38
rs778608358 Chr.3: 151437289-151437289 on Build GRCh38
rs139448889 Chr.3: 151437279-151437279 on Build GRCh38
rs1016929140 Chr.3: 151437415-151437415 on Build GRCh38
rs768415188 Chr.3: 151437414-151437414 on Build GRCh38
rs749050566 Chr.3: 151437406-151437407 on Build GRCh38
rs754285145 Chr.3: 151437232-151437234 on Build GRCh38
rs750121143 Chr.3: 151437251-151437252 on Build GRCh38
rs762262818 Chr.3: 151437255-151437255 on Build GRCh38
rs758246100 Chr.3: 151437359-151437359 on Build GRCh38
rs758073809 Chr.3: 151437296-151437296 on Build GRCh38
rs151130684 Chr.3: 151437346-151437346 on Build GRCh38
rs764847480 Chr.3: 151437309-151437309 on Build GRCh38
rs145036931 Chr.3: 151437302-151437302 on Build GRCh38
rs964464624 Chr.3: 151437237-151437237 on Build GRCh38
rs759502811 Chr.3: 151437334-151437334 on Build GRCh38
rs757704388 Chr.3: 151437237-151437237 on Build GRCh38
rs750263643 Chr.3: 151437355-151437355 on Build GRCh38
rs760604211 Chr.3: 151437212-151437215 on Build GRCh38
rs182380302 Chr.3: 151437250-151437250 on Build GRCh38
rs766976728 Chr.3: 151437222-151437222 on Build GRCh38
rs925754363 Chr.3: 151437325-151437325 on Build GRCh38
rs751831835 Chr.3: 151437380-151437380 on Build GRCh38
rs773119765 Chr.3: 151437400-151437401 on Build GRCh38
rs938496486 Chr.3: 151437374-151437374 on Build GRCh38
rs781249731 Chr.3: 151437372-151437373 on Build GRCh38
rs141148204 Chr.3: 151437381-151437381 on Build GRCh38
rs112300737 Chr.3: 151437256-151437256 on Build GRCh38
rs779779499 Chr.3: 151437362-151437362 on Build GRCh38
rs748062082 Chr.3: 151437224-151437225 on Build GRCh38
rs775743050 Chr.3: 151437212-151437213 on Build GRCh38
rs747196885 Chr.3: 151437301-151437301 on Build GRCh38
rs769341119 Chr.3: 151437278-151437279 on Build GRCh38
rs757033637 Chr.3: 151437241-151437241 on Build GRCh38
rs772304441 Chr.3: 151437247-151437247 on Build GRCh38
rs769752837 Chr.3: 151437382-151437383 on Build GRCh38
rs753363334 Chr.3: 151437353-151437353 on Build GRCh38
rs775610704 Chr.3: 151437248-151437248 on Build GRCh38
rs767186809 Chr.3: 151437266-151437266 on Build GRCh38
rs146894591 Chr.3: 151437382-151437382 on Build GRCh38
rs533904682 Chr.3: 151437410-151437410 on Build GRCh38
rs756176928 Chr.3: 151437229-151437229 on Build GRCh38
rs867084352 Chr.3: 151437341-151437341 on Build GRCh38
rs770332027 Chr.3: 151437315-151437315 on Build GRCh38
rs764003846 Chr.3: 151437287-151437287 on Build GRCh38
rs984102189 Chr.3: 151437337-151437337 on Build GRCh38
rs111254790 Chr.3: 151437307-151437307 on Build GRCh38
rs370875040 Chr.3: 151437332-151437332 on Build GRCh38
rs771354299 Chr.3: 151437333-151437333 on Build GRCh38
rs761721524 Chr.3: 151437354-151437354 on Build GRCh38
rs773595291 Chr.3: 151437263-151437263 on Build GRCh38
rs779645184 Chr.3: 151437296-151437297 on Build GRCh38
rs374159434 Chr.3: 151437225-151437225 on Build GRCh38
rs775313750 Chr.3: 151437335-151437335 on Build GRCh38
rs373513528 Chr.3: 151437243-151437243 on Build GRCh38
rs954661445 Chr.3: 151437282-151437282 on Build GRCh38
rs111468413 Chr.3: 151437213-151437213 on Build GRCh38
rs777158027 Chr.3: 151437254-151437254 on Build GRCh38
rs762172767 Chr.3: 151437210-151437210 on Build GRCh38
rs754391210 Chr.3: 151437367-151437368 on Build GRCh38
rs780262265 Chr.3: 151437298-151437298 on Build GRCh38
rs141741834 Chr.3: 151437409-151437409 on Build GRCh38
rs751194629 Chr.3: 151437215-151437215 on Build GRCh38
rs755687026 Chr.3: 151437273-151437273 on Build GRCh38
rs759180744 Chr.3: 151437264-151437264 on Build GRCh38
rs750226083 Chr.3: 151437291-151437291 on Build GRCh38
rs138165000 Chr.3: 151437344-151437344 on Build GRCh38
rs377460296 Chr.3: 151437267-151437267 on Build GRCh38
rs749171295 Chr.3: 151437240-151437240 on Build GRCh38
rs768658512 Chr.3: 151437253-151437253 on Build GRCh38
rs773508865 Chr.3: 151437320-151437320 on Build GRCh38
rs548116940 Chr.3: 151437412-151437412 on Build GRCh38
rs781227634 Chr.3: 151437305-151437305 on Build GRCh38
rs1021942646 Chr.3: 151437400-151437400 on Build GRCh38
rs765552954 Chr.3: 151437250-151437251 on Build GRCh38
rs746121345 Chr.3: 151437244-151437244 on Build GRCh38
rs769706595 Chr.3: 151437383-151437383 on Build GRCh38
rs778347032 Chr.3: 151437408-151437408 on Build GRCh38
rs746536968 Chr.3: 151437355-151437356 on Build GRCh38
rs753847419 Chr.3: 151437280-151437280 on Build GRCh38
rs777864278 Chr.3: 151437236-151437236 on Build GRCh38

View Full Product Details