Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63168352-63168623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6867496 c.7485G>A p.Q2495Q Chr.10: 63168483-63168483 on Build GRCh38
6482461 c.6717G>A p.Q2239Q Chr.10: 63168540-63168540 on Build GRCh38
6482460 c.7428G>A p.Q2476Q Chr.10: 63168540-63168540 on Build GRCh38
231213 c.7416C>A p.H2472Q Chr.10: 63168552-63168552 on Build GRCh38
304330 c.6784C>A p.L2262I Chr.10: 63168473-63168473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751861560 Chr.10: 63168418-63168418 on Build GRCh38
rs372033085 Chr.10: 63168491-63168491 on Build GRCh38
rs746631132 Chr.10: 63168566-63168566 on Build GRCh38
rs529698959 Chr.10: 63168465-63168465 on Build GRCh38
rs768345019 Chr.10: 63168576-63168576 on Build GRCh38
rs200089221 Chr.10: 63168587-63168587 on Build GRCh38
rs779638094 Chr.10: 63168545-63168545 on Build GRCh38
rs766664052 Chr.10: 63168412-63168412 on Build GRCh38
rs368646956 Chr.10: 63168409-63168409 on Build GRCh38
rs549553282 Chr.10: 63168493-63168493 on Build GRCh38
rs758211612 Chr.10: 63168535-63168535 on Build GRCh38
rs140275881 Chr.10: 63168406-63168406 on Build GRCh38
rs755292204 Chr.10: 63168449-63168449 on Build GRCh38
rs747701384 Chr.10: 63168594-63168594 on Build GRCh38
rs532878838 Chr.10: 63168473-63168473 on Build GRCh38
rs959708656 Chr.10: 63168387-63168387 on Build GRCh38
rs779521920 Chr.10: 63168584-63168585 on Build GRCh38
rs752882287 Chr.10: 63168480-63168480 on Build GRCh38
rs749097662 Chr.10: 63168388-63168388 on Build GRCh38
rs946086198 Chr.10: 63168415-63168415 on Build GRCh38
rs745591716 Chr.10: 63168522-63168522 on Build GRCh38

View Full Product Details