Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68224980-68225241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL26
Entrez Gene ID
55801
Gene Name
interleukin 26
Gene Aliases
AK155, IL-26
UniGene
Hs.272350
Gene Chromosome Location
Chr.12: 68201349-68225791 on Build GRCh38
Transcripts
NM_018402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
389066 c.356G>T p.S119I Chr.12: 68225156-68225156 on Build GRCh38
5841780 c.322G>T p.V108L Chr.12: 68225190-68225190 on Build GRCh38
1363844 c.274G>T p.D92Y Chr.12: 68225238-68225238 on Build GRCh38
4954828 c.290T>A p.L97Q Chr.12: 68225222-68225222 on Build GRCh38
378163 c.343A>G p.R115G Chr.12: 68225169-68225169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148742560 Chr.12: 68225173-68225173 on Build GRCh38
rs780593964 Chr.12: 68225175-68225175 on Build GRCh38
rs377625824 Chr.12: 68225126-68225126 on Build GRCh38
rs960762486 Chr.12: 68225050-68225050 on Build GRCh38
rs202164869 Chr.12: 68225156-68225156 on Build GRCh38
rs758938498 Chr.12: 68225174-68225174 on Build GRCh38
rs1049007118 Chr.12: 68225122-68225122 on Build GRCh38
rs542964570 Chr.12: 68225208-68225208 on Build GRCh38
rs772672754 Chr.12: 68225190-68225190 on Build GRCh38
rs1056759129 Chr.12: 68225204-68225204 on Build GRCh38
rs373868127 Chr.12: 68225196-68225196 on Build GRCh38
rs926412937 Chr.12: 68225045-68225045 on Build GRCh38
rs991991534 Chr.12: 68225055-68225055 on Build GRCh38
rs1050429491 Chr.12: 68225082-68225082 on Build GRCh38
rs922603257 Chr.12: 68225052-68225052 on Build GRCh38
rs895467936 Chr.12: 68225216-68225216 on Build GRCh38
rs747325527 Chr.12: 68225176-68225176 on Build GRCh38
rs150012709 Chr.12: 68225135-68225135 on Build GRCh38
rs140471808 Chr.12: 68225089-68225089 on Build GRCh38
rs980462421 Chr.12: 68225040-68225040 on Build GRCh38
rs553468026 Chr.12: 68225104-68225104 on Build GRCh38
rs767349367 Chr.12: 68225130-68225130 on Build GRCh38
rs879266345 Chr.12: 68225101-68225101 on Build GRCh38
rs369981889 Chr.12: 68225166-68225166 on Build GRCh38
rs572123910 Chr.12: 68225127-68225127 on Build GRCh38
rs769498931 Chr.12: 68225183-68225183 on Build GRCh38
rs774220453 Chr.12: 68225116-68225116 on Build GRCh38
rs567721891 Chr.12: 68225053-68225053 on Build GRCh38
rs753967420 Chr.12: 68225162-68225162 on Build GRCh38
rs145221946 Chr.12: 68225160-68225160 on Build GRCh38
rs756360493 Chr.12: 68225141-68225141 on Build GRCh38
rs773889738 Chr.12: 68225218-68225218 on Build GRCh38

View Full Product Details