Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32893397-32893634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPCA
Entrez Gene ID
3208
Gene Name
hippocalcin
Gene Aliases
BDR2, DYT2
UniGene
Hs.632391
Gene Chromosome Location
Chr.1: 32885965-32894646 on Build GRCh38
Transcripts
NM_002143.2 XM_005270792.2 XM_017001118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3865339 c.420G>A p.E140E Chr.1: 32893565-32893565 on Build GRCh38
4186115 c.435delG p.E146fs*>48 Chr.1: 32893580-32893580 on Build GRCh38
2078478 c.441G>T p.K147N Chr.1: 32893586-32893586 on Build GRCh38
7273539 c.472A>T p.T158S Chr.1: 32893617-32893617 on Build GRCh38
6681852 c.408_410delGAA p.K137delK Chr.1: 32893553-32893555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774067129 Chr.1: 32893501-32893501 on Build GRCh38
rs557895597 Chr.1: 32893520-32893520 on Build GRCh38
rs781778640 Chr.1: 32893562-32893562 on Build GRCh38
rs375169113 Chr.1: 32893493-32893493 on Build GRCh38
rs772874379 Chr.1: 32893489-32893489 on Build GRCh38
rs964703426 Chr.1: 32893484-32893484 on Build GRCh38
rs747528254 Chr.1: 32893474-32893474 on Build GRCh38
rs987799748 Chr.1: 32893453-32893453 on Build GRCh38
rs774052115 Chr.1: 32893592-32893592 on Build GRCh38
rs955892975 Chr.1: 32893464-32893464 on Build GRCh38
rs200141644 Chr.1: 32893508-32893508 on Build GRCh38
rs747269664 Chr.1: 32893559-32893559 on Build GRCh38
rs762326492 Chr.1: 32893487-32893487 on Build GRCh38
rs762547556 Chr.1: 32893495-32893495 on Build GRCh38
rs767512930 Chr.1: 32893505-32893505 on Build GRCh38
rs766169582 Chr.1: 32893500-32893500 on Build GRCh38
rs770427605 Chr.1: 32893585-32893585 on Build GRCh38
rs769444903 Chr.1: 32893483-32893483 on Build GRCh38
rs373435512 Chr.1: 32893521-32893521 on Build GRCh38
rs748987737 Chr.1: 32893584-32893584 on Build GRCh38
rs748641503 Chr.1: 32893563-32893563 on Build GRCh38
rs376349097 Chr.1: 32893548-32893548 on Build GRCh38
rs755388440 Chr.1: 32893561-32893561 on Build GRCh38
rs769465555 Chr.1: 32893573-32893573 on Build GRCh38
rs377608024 Chr.1: 32893443-32893443 on Build GRCh38
rs954609865 Chr.1: 32893452-32893452 on Build GRCh38
rs910505105 Chr.1: 32893471-32893471 on Build GRCh38
rs759526373 Chr.1: 32893504-32893504 on Build GRCh38
rs761342009 Chr.1: 32893482-32893482 on Build GRCh38
rs905773304 Chr.1: 32893425-32893425 on Build GRCh38
rs776277238 Chr.1: 32893478-32893478 on Build GRCh38
rs772554177 Chr.1: 32893576-32893576 on Build GRCh38
rs1000531329 Chr.1: 32893441-32893441 on Build GRCh38
rs140440243 Chr.1: 32893557-32893557 on Build GRCh38

View Full Product Details