Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132749556-132749817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5A
Entrez Gene ID
3632
Gene Name
inositol polyphosphate-5-phosphatase A
Gene Aliases
5PTASE
UniGene
Hs.523360
Gene Chromosome Location
Chr.10: 132537779-132783480 on Build GRCh38
Transcripts
NM_001321042.1 NM_005539.4 XM_017016204.1 XM_017016205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916377 c.849G>T p.K283N Chr.10: 132749791-132749791 on Build GRCh38
2023549 c.774C>T p.A258A Chr.10: 132749558-132749558 on Build GRCh38
916375 c.820G>A p.D274N Chr.10: 132749604-132749604 on Build GRCh38
1177141 c.784G>T p.E262* Chr.10: 132749568-132749568 on Build GRCh38
3747154 c.828+6C>T p.? Chr.10: 132749618-132749618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038841470 Chr.10: 132749714-132749714 on Build GRCh38
rs983364721 Chr.10: 132749683-132749683 on Build GRCh38
rs755197376 Chr.10: 132749748-132749748 on Build GRCh38
rs776310280 Chr.10: 132749619-132749619 on Build GRCh38
rs950704723 Chr.10: 132749668-132749668 on Build GRCh38
rs765177916 Chr.10: 132749625-132749625 on Build GRCh38
rs147362550 Chr.10: 132749604-132749604 on Build GRCh38
rs200837473 Chr.10: 132749607-132749607 on Build GRCh38
rs368760548 Chr.10: 132749622-132749622 on Build GRCh38
rs918559902 Chr.10: 132749666-132749666 on Build GRCh38
rs41302997 Chr.10: 132749696-132749696 on Build GRCh38
rs147310419 Chr.10: 132749603-132749603 on Build GRCh38
rs752531674 Chr.10: 132749626-132749626 on Build GRCh38
rs751472060 Chr.10: 132749731-132749731 on Build GRCh38
rs774667640 Chr.10: 132749599-132749599 on Build GRCh38
rs758724246 Chr.10: 132749755-132749755 on Build GRCh38
rs376682292 Chr.10: 132749651-132749651 on Build GRCh38
rs751163648 Chr.10: 132749757-132749758 on Build GRCh38
rs759102705 Chr.10: 132749702-132749702 on Build GRCh38
rs753848782 Chr.10: 132749743-132749743 on Build GRCh38
rs568247188 Chr.10: 132749662-132749662 on Build GRCh38
rs766929246 Chr.10: 132749684-132749684 on Build GRCh38
rs111561600 Chr.10: 132749646-132749646 on Build GRCh38
rs376203197 Chr.10: 132749753-132749753 on Build GRCh38
rs201098272 Chr.10: 132749643-132749643 on Build GRCh38
rs181742238 Chr.10: 132749647-132749647 on Build GRCh38
rs139448022 Chr.10: 132749637-132749637 on Build GRCh38
rs3829130 Chr.10: 132749618-132749618 on Build GRCh38
rs770517998 Chr.10: 132749659-132749659 on Build GRCh38
rs143990293 Chr.10: 132749597-132749597 on Build GRCh38
rs372793753 Chr.10: 132749608-132749608 on Build GRCh38
rs547626612 Chr.10: 132749720-132749720 on Build GRCh38
rs1031033515 Chr.10: 132749623-132749623 on Build GRCh38
rs747603691 Chr.10: 132749771-132749771 on Build GRCh38
rs760176056 Chr.10: 132749617-132749617 on Build GRCh38
rs750479854 Chr.10: 132749638-132749638 on Build GRCh38
rs762730484 Chr.10: 132749617-132749618 on Build GRCh38
rs201962186 Chr.10: 132749641-132749641 on Build GRCh38
rs201167385 Chr.10: 132749590-132749590 on Build GRCh38
rs201214747 Chr.10: 132749596-132749596 on Build GRCh38
rs762776248 Chr.10: 132749725-132749725 on Build GRCh38
rs185486477 Chr.10: 132749656-132749656 on Build GRCh38
rs766528692 Chr.10: 132749735-132749735 on Build GRCh38
rs879144960 Chr.10: 132749712-132749712 on Build GRCh38
rs763638788 Chr.10: 132749634-132749655 on Build GRCh38
rs763415049 Chr.10: 132749678-132749678 on Build GRCh38
rs918166654 Chr.10: 132749691-132749691 on Build GRCh38
rs775426199 Chr.10: 132749721-132749721 on Build GRCh38
rs371253241 Chr.10: 132749655-132749655 on Build GRCh38
rs758472291 Chr.10: 132749627-132749627 on Build GRCh38
rs748536319 Chr.10: 132749754-132749754 on Build GRCh38
rs138441487 Chr.10: 132749583-132749583 on Build GRCh38
rs767124935 Chr.10: 132749764-132749764 on Build GRCh38
rs377725435 Chr.10: 132749642-132749642 on Build GRCh38
rs190355369 Chr.10: 132749728-132749728 on Build GRCh38
rs375440993 Chr.10: 132749589-132749589 on Build GRCh38

View Full Product Details