Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35635326-35635581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERPUD2
Entrez Gene ID
64224
Gene Name
HERPUD family member 2
Gene Aliases
-
UniGene
Hs.643051
Gene Chromosome Location
Chr.7: 35632659-35695187 on Build GRCh38
Transcripts
NM_022373.4 XM_006715765.1 XM_011515473.1 XM_017012506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6248743 c.733C>A p.L245I Chr.7: 35635343-35635343 on Build GRCh38
3995564 c.722C>A p.P241Q Chr.7: 35635354-35635354 on Build GRCh38
5634403 c.647C>T p.S216L Chr.7: 35635429-35635429 on Build GRCh38
340794 c.636T>A p.A212A Chr.7: 35635440-35635440 on Build GRCh38
7114605 c.685C>G p.L229V Chr.7: 35635391-35635391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760838074 Chr.7: 35635361-35635361 on Build GRCh38
rs994379293 Chr.7: 35635521-35635521 on Build GRCh38
rs765078899 Chr.7: 35635415-35635415 on Build GRCh38
rs763901803 Chr.7: 35635478-35635478 on Build GRCh38
rs752639535 Chr.7: 35635426-35635426 on Build GRCh38
rs770529059 Chr.7: 35635491-35635491 on Build GRCh38
rs756829494 Chr.7: 35635484-35635484 on Build GRCh38
rs772577226 Chr.7: 35635359-35635359 on Build GRCh38
rs776546948 Chr.7: 35635398-35635398 on Build GRCh38
rs201820568 Chr.7: 35635407-35635407 on Build GRCh38
rs780948572 Chr.7: 35635491-35635491 on Build GRCh38
rs536626540 Chr.7: 35635394-35635394 on Build GRCh38
rs758135857 Chr.7: 35635431-35635431 on Build GRCh38
rs773357768 Chr.7: 35635362-35635362 on Build GRCh38
rs773518701 Chr.7: 35635360-35635360 on Build GRCh38
rs751173240 Chr.7: 35635481-35635481 on Build GRCh38
rs947768763 Chr.7: 35635405-35635405 on Build GRCh38
rs746682433 Chr.7: 35635490-35635491 on Build GRCh38
rs898658314 Chr.7: 35635503-35635503 on Build GRCh38
rs772682821 Chr.7: 35635366-35635377 on Build GRCh38
rs542711530 Chr.7: 35635515-35635515 on Build GRCh38
rs745929522 Chr.7: 35635410-35635410 on Build GRCh38
rs573628381 Chr.7: 35635507-35635507 on Build GRCh38

View Full Product Details