Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1524681-1524948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT140
Entrez Gene ID
9742
Gene Name
intraflagellar transport 140
Gene Aliases
MZSDS, SRTD9, WDTC2, c305C8.4, c380F5.1, gs114
UniGene
Hs.389438
Gene Chromosome Location
Chr.16: 1510427-1612108 on Build GRCh38
Transcripts
NM_014714.3 XM_006720990.3 XM_006720991.3 XM_006720992.3 XM_011522766.2 XM_011522767.2 XM_017023910.1 XM_017023911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
370772 c.2965C>T p.R989C Chr.16: 1524816-1524816 on Build GRCh38
4058544 c.2875C>T p.R959W Chr.16: 1524906-1524906 on Build GRCh38
118835 c.2916C>T p.D972D Chr.16: 1524865-1524865 on Build GRCh38
4770096 c.2979C>T p.F993F Chr.16: 1524802-1524802 on Build GRCh38
5342692 c.2903A>G p.Q968R Chr.16: 1524878-1524878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537085826 Chr.16: 1524906-1524906 on Build GRCh38
rs756125540 Chr.16: 1524728-1524730 on Build GRCh38
rs148240226 Chr.16: 1524905-1524905 on Build GRCh38
rs760315708 Chr.16: 1524750-1524750 on Build GRCh38
rs147756729 Chr.16: 1524816-1524816 on Build GRCh38
rs201864536 Chr.16: 1524859-1524859 on Build GRCh38
rs765573126 Chr.16: 1524892-1524892 on Build GRCh38
rs761061336 Chr.16: 1524869-1524869 on Build GRCh38
rs769420945 Chr.16: 1524861-1524861 on Build GRCh38
rs1006412072 Chr.16: 1524787-1524787 on Build GRCh38
rs745576178 Chr.16: 1524860-1524860 on Build GRCh38
rs375612616 Chr.16: 1524761-1524761 on Build GRCh38
rs372676892 Chr.16: 1524738-1524738 on Build GRCh38
rs986753439 Chr.16: 1524709-1524709 on Build GRCh38
rs894474570 Chr.16: 1524924-1524924 on Build GRCh38
rs745980432 Chr.16: 1524789-1524789 on Build GRCh38
rs369493147 Chr.16: 1524864-1524864 on Build GRCh38
rs766045677 Chr.16: 1524753-1524753 on Build GRCh38
rs772919008 Chr.16: 1524797-1524797 on Build GRCh38
rs367724835 Chr.16: 1524824-1524824 on Build GRCh38
rs911100501 Chr.16: 1524713-1524713 on Build GRCh38
rs777862012 Chr.16: 1524847-1524847 on Build GRCh38
rs764492842 Chr.16: 1524815-1524815 on Build GRCh38
rs750828167 Chr.16: 1524828-1524828 on Build GRCh38
rs145645423 Chr.16: 1524794-1524794 on Build GRCh38
rs770284479 Chr.16: 1524805-1524805 on Build GRCh38
rs373121582 Chr.16: 1524721-1524721 on Build GRCh38
rs373868663 Chr.16: 1524778-1524778 on Build GRCh38
rs369533072 Chr.16: 1524717-1524717 on Build GRCh38
rs143720412 Chr.16: 1524873-1524873 on Build GRCh38
rs986900353 Chr.16: 1524849-1524849 on Build GRCh38
rs142342085 Chr.16: 1524837-1524837 on Build GRCh38
rs910870360 Chr.16: 1524858-1524858 on Build GRCh38
rs372183503 Chr.16: 1524776-1524776 on Build GRCh38
rs1054312904 Chr.16: 1524728-1524728 on Build GRCh38
rs375635975 Chr.16: 1524811-1524811 on Build GRCh38
rs770497567 Chr.16: 1524851-1524851 on Build GRCh38
rs146277966 Chr.16: 1524846-1524846 on Build GRCh38
rs752903993 Chr.16: 1524893-1524893 on Build GRCh38
rs749121321 Chr.16: 1524730-1524730 on Build GRCh38
rs746546785 Chr.16: 1524800-1524800 on Build GRCh38
rs946595729 Chr.16: 1524777-1524777 on Build GRCh38
rs777383314 Chr.16: 1524900-1524900 on Build GRCh38
rs200390733 Chr.16: 1524772-1524772 on Build GRCh38
rs201150342 Chr.16: 1524895-1524895 on Build GRCh38
rs551125772 Chr.16: 1524773-1524773 on Build GRCh38
rs368150661 Chr.16: 1524762-1524762 on Build GRCh38
rs201097047 Chr.16: 1524747-1524747 on Build GRCh38
rs377725041 Chr.16: 1524754-1524754 on Build GRCh38
rs748046598 Chr.16: 1524744-1524744 on Build GRCh38
rs201139143 Chr.16: 1524743-1524743 on Build GRCh38
rs1027711761 Chr.16: 1524739-1524739 on Build GRCh38
rs753855080 Chr.16: 1524877-1524877 on Build GRCh38
rs113784788 Chr.16: 1524920-1524920 on Build GRCh38
rs376610727 Chr.16: 1524722-1524722 on Build GRCh38
rs777513393 Chr.16: 1524736-1524737 on Build GRCh38
rs1053132029 Chr.16: 1524803-1524803 on Build GRCh38
rs780823837 Chr.16: 1524915-1524915 on Build GRCh38
rs759898401 Chr.16: 1524885-1524885 on Build GRCh38
rs2235640 Chr.16: 1524862-1524862 on Build GRCh38
rs766316995 Chr.16: 1524836-1524836 on Build GRCh38
rs779711939 Chr.16: 1524921-1524921 on Build GRCh38
rs995478287 Chr.16: 1524840-1524840 on Build GRCh38
rs768024276 Chr.16: 1524826-1524826 on Build GRCh38
rs752529928 Chr.16: 1524844-1524844 on Build GRCh38
rs750488202 Chr.16: 1524770-1524770 on Build GRCh38
rs778921745 Chr.16: 1524841-1524841 on Build GRCh38
rs542019042 Chr.16: 1524759-1524759 on Build GRCh38
rs78178397 Chr.16: 1524793-1524793 on Build GRCh38
rs148945612 Chr.16: 1524865-1524865 on Build GRCh38
rs760839341 Chr.16: 1524829-1524829 on Build GRCh38
rs757358956 Chr.16: 1524720-1524720 on Build GRCh38
rs779768890 Chr.16: 1524727-1524727 on Build GRCh38
rs375561732 Chr.16: 1524848-1524848 on Build GRCh38

View Full Product Details