Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52799690-52799939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH3
Entrez Gene ID
3699
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
H3P, ITI-HC3, SHAP
UniGene
Hs.76716
Gene Chromosome Location
Chr.3: 52794768-52809009 on Build GRCh38
Transcripts
NM_002217.3 XM_005265105.4 XM_017006356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532879 c.979G>A p.G327R Chr.3: 52799825-52799825 on Build GRCh38
5532878 c.979G>A p.G327R Chr.3: 52799825-52799825 on Build GRCh38
1255062 c.919C>A p.L307I Chr.3: 52799765-52799765 on Build GRCh38
7414522 c.1059C>A p.S353R Chr.3: 52799905-52799905 on Build GRCh38
1255061 c.919C>A p.L307I Chr.3: 52799765-52799765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759134530 Chr.3: 52799848-52799848 on Build GRCh38
rs749234695 Chr.3: 52799817-52799817 on Build GRCh38
rs373955614 Chr.3: 52799885-52799885 on Build GRCh38
rs200979613 Chr.3: 52799896-52799896 on Build GRCh38
rs768686512 Chr.3: 52799889-52799889 on Build GRCh38
rs370112904 Chr.3: 52799818-52799818 on Build GRCh38
rs374644581 Chr.3: 52799790-52799790 on Build GRCh38
rs937822472 Chr.3: 52799813-52799813 on Build GRCh38
rs776658926 Chr.3: 52799916-52799916 on Build GRCh38
rs754345398 Chr.3: 52799757-52799757 on Build GRCh38
rs74587399 Chr.3: 52799869-52799869 on Build GRCh38
rs767742844 Chr.3: 52799826-52799826 on Build GRCh38
rs1042500561 Chr.3: 52799868-52799868 on Build GRCh38
rs544210438 Chr.3: 52799834-52799834 on Build GRCh38
rs753973123 Chr.3: 52799861-52799861 on Build GRCh38
rs778545922 Chr.3: 52799845-52799846 on Build GRCh38
rs369039370 Chr.3: 52799715-52799716 on Build GRCh38
rs377554371 Chr.3: 52799892-52799892 on Build GRCh38
rs779319143 Chr.3: 52799763-52799763 on Build GRCh38
rs35271262 Chr.3: 52799865-52799865 on Build GRCh38
rs568928822 Chr.3: 52799902-52799902 on Build GRCh38
rs190237910 Chr.3: 52799908-52799908 on Build GRCh38
rs769444762 Chr.3: 52799803-52799803 on Build GRCh38
rs186486873 Chr.3: 52799863-52799863 on Build GRCh38
rs758711384 Chr.3: 52799867-52799867 on Build GRCh38
rs774081410 Chr.3: 52799721-52799721 on Build GRCh38
rs755542139 Chr.3: 52799761-52799761 on Build GRCh38
rs3617 Chr.3: 52799789-52799789 on Build GRCh38
rs985103353 Chr.3: 52799771-52799771 on Build GRCh38
rs767686015 Chr.3: 52799734-52799734 on Build GRCh38
rs748860080 Chr.3: 52799886-52799886 on Build GRCh38
rs756902170 Chr.3: 52799787-52799787 on Build GRCh38
rs773316727 Chr.3: 52799830-52799830 on Build GRCh38
rs181053134 Chr.3: 52799742-52799742 on Build GRCh38
rs200095865 Chr.3: 52799870-52799870 on Build GRCh38
rs753066152 Chr.3: 52799866-52799866 on Build GRCh38
rs774177038 Chr.3: 52799819-52799819 on Build GRCh38
rs757010954 Chr.3: 52799711-52799712 on Build GRCh38
rs766811546 Chr.3: 52799750-52799750 on Build GRCh38
rs775501840 Chr.3: 52799806-52799806 on Build GRCh38
rs761558025 Chr.3: 52799824-52799824 on Build GRCh38
rs745441762 Chr.3: 52799891-52799892 on Build GRCh38
rs377601188 Chr.3: 52799722-52799722 on Build GRCh38
rs750487090 Chr.3: 52799740-52799740 on Build GRCh38
rs766439437 Chr.3: 52799854-52799854 on Build GRCh38
rs201984888 Chr.3: 52799893-52799893 on Build GRCh38

View Full Product Details