Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 45262023-45262276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN1
Entrez Gene ID
348980
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Gene Aliases
BCNG-1, BCNG1, EIEE24, HAC-2
UniGene
Hs.353176
Gene Chromosome Location
Chr.5: 45254950-45696118 on Build GRCh38
Transcripts
NM_021072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6200880 c.2471G>A p.G824E Chr.5: 45262123-45262123 on Build GRCh38
1254000 c.2565A>G p.G855G Chr.5: 45262029-45262029 on Build GRCh38
3920130 c.2523C>T p.L841L Chr.5: 45262071-45262071 on Build GRCh38
738681 c.2460G>C p.T820T Chr.5: 45262134-45262134 on Build GRCh38
5667701 c.2543G>A p.G848E Chr.5: 45262051-45262051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888196323 Chr.5: 45262070-45262070 on Build GRCh38
rs770074008 Chr.5: 45262160-45262160 on Build GRCh38
rs758609806 Chr.5: 45262233-45262233 on Build GRCh38
rs777158203 Chr.5: 45262156-45262156 on Build GRCh38
rs772076370 Chr.5: 45262134-45262134 on Build GRCh38
rs575150344 Chr.5: 45262251-45262251 on Build GRCh38
rs753637793 Chr.5: 45262119-45262119 on Build GRCh38
rs752908921 Chr.5: 45262223-45262223 on Build GRCh38
rs1046832984 Chr.5: 45262067-45262067 on Build GRCh38
rs948061807 Chr.5: 45262230-45262230 on Build GRCh38
rs761336904 Chr.5: 45262092-45262092 on Build GRCh38
rs368989823 Chr.5: 45262087-45262087 on Build GRCh38
rs140758934 Chr.5: 45262219-45262219 on Build GRCh38
rs776970041 Chr.5: 45262072-45262072 on Build GRCh38
rs998527315 Chr.5: 45262144-45262144 on Build GRCh38
rs759882440 Chr.5: 45262082-45262082 on Build GRCh38
rs756131996 Chr.5: 45262102-45262102 on Build GRCh38
rs772068771 Chr.5: 45262044-45262044 on Build GRCh38
rs727503956 Chr.5: 45262166-45262166 on Build GRCh38
rs572698864 Chr.5: 45262054-45262054 on Build GRCh38
rs771230431 Chr.5: 45262060-45262060 on Build GRCh38
rs767166082 Chr.5: 45262095-45262095 on Build GRCh38
rs746392507 Chr.5: 45262254-45262254 on Build GRCh38
rs778039525 Chr.5: 45262241-45262241 on Build GRCh38
rs967232218 Chr.5: 45262162-45262162 on Build GRCh38
rs751215219 Chr.5: 45262090-45262090 on Build GRCh38
rs150936707 Chr.5: 45262204-45262204 on Build GRCh38
rs780209007 Chr.5: 45262103-45262103 on Build GRCh38
rs754764304 Chr.5: 45262212-45262212 on Build GRCh38
rs267600644 Chr.5: 45262098-45262098 on Build GRCh38
rs760303020 Chr.5: 45262197-45262197 on Build GRCh38
rs375360013 Chr.5: 45262149-45262149 on Build GRCh38
rs371602396 Chr.5: 45262132-45262132 on Build GRCh38
rs774013410 Chr.5: 45262167-45262167 on Build GRCh38
rs759920350 Chr.5: 45262157-45262157 on Build GRCh38
rs958111537 Chr.5: 45262200-45262200 on Build GRCh38
rs1035068089 Chr.5: 45262175-45262175 on Build GRCh38
rs199887416 Chr.5: 45262099-45262099 on Build GRCh38
rs766397326 Chr.5: 45262198-45262198 on Build GRCh38
rs779149243 Chr.5: 45262128-45262128 on Build GRCh38
rs781686163 Chr.5: 45262253-45262253 on Build GRCh38
rs1018363057 Chr.5: 45262131-45262131 on Build GRCh38
rs777848314 Chr.5: 45262138-45262138 on Build GRCh38
rs767523812 Chr.5: 45262247-45262247 on Build GRCh38
rs779990153 Chr.5: 45262125-45262125 on Build GRCh38
rs749939446 Chr.5: 45262188-45262188 on Build GRCh38
rs767078014 Chr.5: 45262172-45262172 on Build GRCh38
rs761374276 Chr.5: 45262171-45262171 on Build GRCh38
rs747153982 Chr.5: 45262140-45262140 on Build GRCh38
rs142485423 Chr.5: 45262125-45262125 on Build GRCh38
rs1003463768 Chr.5: 45262150-45262150 on Build GRCh38

View Full Product Details