Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176169439-176169682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD3
Entrez Gene ID
3232
Gene Name
homeobox D3
Gene Aliases
HOX1D, HOX4, HOX4A, Hox-4.1
UniGene
Hs.93574
Gene Chromosome Location
Chr.2: 176136921-176173102 on Build GRCh38
Transcripts
NM_006898.4 XM_005246509.3 XM_005246510.4 XM_005246511.3 XM_005246513.4 XM_006712477.2 XM_011511065.2 XM_011511066.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305874 c.393C>A p.P131P Chr.2: 176169507-176169507 on Build GRCh38
571251 c.429C>A p.P143P Chr.2: 176169543-176169543 on Build GRCh38
4492626 c.400C>T p.P134S Chr.2: 176169514-176169514 on Build GRCh38
5685310 c.505C>A p.Q169K Chr.2: 176169619-176169619 on Build GRCh38
7208264 c.407G>T p.G136V Chr.2: 176169521-176169521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755084410 Chr.2: 176169521-176169521 on Build GRCh38
rs544638987 Chr.2: 176169663-176169663 on Build GRCh38
rs371779084 Chr.2: 176169588-176169588 on Build GRCh38
rs775027801 Chr.2: 176169591-176169591 on Build GRCh38
rs763764680 Chr.2: 176169618-176169618 on Build GRCh38
rs747438109 Chr.2: 176169478-176169478 on Build GRCh38
rs1051932 Chr.2: 176169500-176169500 on Build GRCh38
rs773686530 Chr.2: 176169472-176169472 on Build GRCh38
rs749119115 Chr.2: 176169547-176169547 on Build GRCh38
rs868358435 Chr.2: 176169655-176169655 on Build GRCh38
rs992437843 Chr.2: 176169534-176169534 on Build GRCh38
rs868071726 Chr.2: 176169598-176169598 on Build GRCh38
rs768745305 Chr.2: 176169564-176169564 on Build GRCh38
rs751531967 Chr.2: 176169498-176169498 on Build GRCh38
rs774350237 Chr.2: 176169569-176169569 on Build GRCh38
rs140883088 Chr.2: 176169485-176169485 on Build GRCh38
rs74698314 Chr.2: 176169488-176169488 on Build GRCh38
rs773779197 Chr.2: 176169582-176169584 on Build GRCh38
rs199953096 Chr.2: 176169536-176169536 on Build GRCh38
rs756448504 Chr.2: 176169531-176169531 on Build GRCh38
rs959740570 Chr.2: 176169512-176169512 on Build GRCh38
rs936826310 Chr.2: 176169660-176169660 on Build GRCh38
rs925480876 Chr.2: 176169639-176169639 on Build GRCh38
rs773514176 Chr.2: 176169626-176169626 on Build GRCh38
rs905235517 Chr.2: 176169489-176169489 on Build GRCh38
rs760287977 Chr.2: 176169617-176169617 on Build GRCh38
rs376541904 Chr.2: 176169636-176169636 on Build GRCh38
rs766927340 Chr.2: 176169494-176169494 on Build GRCh38
rs368322060 Chr.2: 176169479-176169479 on Build GRCh38
rs749208307 Chr.2: 176169464-176169464 on Build GRCh38
rs748361499 Chr.2: 176169530-176169530 on Build GRCh38
rs766726340 Chr.2: 176169665-176169665 on Build GRCh38
rs781191666 Chr.2: 176169523-176169523 on Build GRCh38
rs765563135 Chr.2: 176169486-176169486 on Build GRCh38
rs190587708 Chr.2: 176169473-176169473 on Build GRCh38
rs770258194 Chr.2: 176169460-176169462 on Build GRCh38
rs771597443 Chr.2: 176169590-176169590 on Build GRCh38
rs375537887 Chr.2: 176169471-176169471 on Build GRCh38
rs770715960 Chr.2: 176169557-176169557 on Build GRCh38

View Full Product Details