Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68310905-68311164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
UniGene
Hs.436416
Gene Chromosome Location
Chr.15: 68297433-68432312 on Build GRCh38
Transcripts
NM_001004439.1 XM_005254228.3 XM_011521363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2257875 c.3158G>A p.R1053H Chr.15: 68311010-68311010 on Build GRCh38
4693226 c.3164C>T p.A1055V Chr.15: 68311004-68311004 on Build GRCh38
4756904 c.3089C>T p.A1030V Chr.15: 68311079-68311079 on Build GRCh38
701403 c.3129C>T p.Y1043Y Chr.15: 68311039-68311039 on Build GRCh38
84295 c.3173T>C p.L1058P Chr.15: 68310995-68310995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036978061 Chr.15: 68310942-68310942 on Build GRCh38
rs752681119 Chr.15: 68310940-68310940 on Build GRCh38
rs547310975 Chr.15: 68311141-68311141 on Build GRCh38
rs201394870 Chr.15: 68311054-68311054 on Build GRCh38
rs754049231 Chr.15: 68311093-68311093 on Build GRCh38
rs909256804 Chr.15: 68310936-68310936 on Build GRCh38
rs370829589 Chr.15: 68311072-68311072 on Build GRCh38
rs986095858 Chr.15: 68311033-68311033 on Build GRCh38
rs745745998 Chr.15: 68310964-68310964 on Build GRCh38
rs768468306 Chr.15: 68311053-68311053 on Build GRCh38
rs909209671 Chr.15: 68311046-68311046 on Build GRCh38
rs777950935 Chr.15: 68311024-68311024 on Build GRCh38
rs758216935 Chr.15: 68311111-68311111 on Build GRCh38
rs372699459 Chr.15: 68310973-68310973 on Build GRCh38
rs376190131 Chr.15: 68311051-68311051 on Build GRCh38
rs778794265 Chr.15: 68311101-68311101 on Build GRCh38
rs141995773 Chr.15: 68311098-68311099 on Build GRCh38
rs146663204 Chr.15: 68311007-68311007 on Build GRCh38
rs752004974 Chr.15: 68311008-68311008 on Build GRCh38
rs771754102 Chr.15: 68310965-68310965 on Build GRCh38
rs770905276 Chr.15: 68310947-68310947 on Build GRCh38
rs199548438 Chr.15: 68311011-68311011 on Build GRCh38
rs763726144 Chr.15: 68310975-68310975 on Build GRCh38
rs781049504 Chr.15: 68311123-68311123 on Build GRCh38
rs896175831 Chr.15: 68310958-68310958 on Build GRCh38
rs879160778 Chr.15: 68311038-68311038 on Build GRCh38
rs948968309 Chr.15: 68311139-68311139 on Build GRCh38
rs919415034 Chr.15: 68310946-68310946 on Build GRCh38
rs775104290 Chr.15: 68311047-68311047 on Build GRCh38
rs1057357959 Chr.15: 68310948-68310948 on Build GRCh38
rs931940197 Chr.15: 68310930-68310930 on Build GRCh38
rs761380839 Chr.15: 68311000-68311000 on Build GRCh38
rs764637683 Chr.15: 68311067-68311067 on Build GRCh38
rs113860589 Chr.15: 68310970-68310970 on Build GRCh38
rs758007495 Chr.15: 68311124-68311124 on Build GRCh38
rs1044747148 Chr.15: 68311140-68311140 on Build GRCh38
rs200056013 Chr.15: 68311037-68311037 on Build GRCh38
rs759923280 Chr.15: 68311073-68311073 on Build GRCh38
rs143218415 Chr.15: 68310931-68310931 on Build GRCh38
rs372902171 Chr.15: 68311031-68311031 on Build GRCh38
rs976006072 Chr.15: 68310927-68310927 on Build GRCh38
rs756439955 Chr.15: 68311079-68311079 on Build GRCh38
rs923292661 Chr.15: 68310929-68310929 on Build GRCh38
rs757328117 Chr.15: 68311030-68311030 on Build GRCh38
rs1031273222 Chr.15: 68310999-68310999 on Build GRCh38
rs765797312 Chr.15: 68311077-68311077 on Build GRCh38
rs370182803 Chr.15: 68311010-68311010 on Build GRCh38
rs551214432 Chr.15: 68310976-68310976 on Build GRCh38
rs749413845 Chr.15: 68311027-68311027 on Build GRCh38
rs749896664 Chr.15: 68311097-68311098 on Build GRCh38
rs761480919 Chr.15: 68311060-68311060 on Build GRCh38
rs745845970 Chr.15: 68311108-68311108 on Build GRCh38
rs369142527 Chr.15: 68311021-68311021 on Build GRCh38
rs1013002474 Chr.15: 68310979-68310979 on Build GRCh38
rs377279143 Chr.15: 68310943-68310943 on Build GRCh38
rs780015298 Chr.15: 68311113-68311113 on Build GRCh38
rs774128492 Chr.15: 68310955-68310955 on Build GRCh38
rs762480298 Chr.15: 68310956-68310956 on Build GRCh38
rs753109919 Chr.15: 68311078-68311078 on Build GRCh38
rs757374689 Chr.15: 68311095-68311095 on Build GRCh38
rs201838249 Chr.15: 68311104-68311104 on Build GRCh38
rs200822139 Chr.15: 68311080-68311080 on Build GRCh38
rs371538230 Chr.15: 68311127-68311127 on Build GRCh38
rs752996913 Chr.15: 68311014-68311014 on Build GRCh38
rs746836825 Chr.15: 68311114-68311114 on Build GRCh38

View Full Product Details