Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19258322-19258587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN4
Entrez Gene ID
404037
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
BRAL2
UniGene
Hs.367829
Gene Chromosome Location
Chr.19: 19255641-19262787 on Build GRCh38
Transcripts
NM_023002.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
711021 c.784T>C p.Y262H Chr.19: 19258556-19258556 on Build GRCh38
4409566 c.816G>A p.P272P Chr.19: 19258524-19258524 on Build GRCh38
5470941 c.789C>T p.D263D Chr.19: 19258551-19258551 on Build GRCh38
5464436 c.817+9G>T p.? Chr.19: 19258514-19258514 on Build GRCh38
6299862 c.817+5G>T p.? Chr.19: 19258518-19258518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184860421 Chr.19: 19258402-19258402 on Build GRCh38
rs554477011 Chr.19: 19258349-19258349 on Build GRCh38
rs1021499888 Chr.19: 19258488-19258488 on Build GRCh38
rs569678736 Chr.19: 19258448-19258448 on Build GRCh38
rs1035752352 Chr.19: 19258467-19258467 on Build GRCh38
rs141246392 Chr.19: 19258441-19258441 on Build GRCh38
rs907649409 Chr.19: 19258377-19258377 on Build GRCh38
rs750722977 Chr.19: 19258530-19258530 on Build GRCh38
rs541996838 Chr.19: 19258470-19258470 on Build GRCh38
rs968960492 Chr.19: 19258494-19258494 on Build GRCh38
rs760946446 Chr.19: 19258531-19258531 on Build GRCh38
rs990220651 Chr.19: 19258415-19258415 on Build GRCh38
rs545136731 Chr.19: 19258514-19258514 on Build GRCh38
rs1055570433 Chr.19: 19258457-19258457 on Build GRCh38
rs987152997 Chr.19: 19258354-19258354 on Build GRCh38
rs749975165 Chr.19: 19258394-19258394 on Build GRCh38
rs779091799 Chr.19: 19258563-19258563 on Build GRCh38
rs767885551 Chr.19: 19258524-19258524 on Build GRCh38
rs144834688 Chr.19: 19258551-19258551 on Build GRCh38
rs1037230875 Chr.19: 19258400-19258400 on Build GRCh38
rs781137725 Chr.19: 19258566-19258566 on Build GRCh38
rs778025794 Chr.19: 19258558-19258558 on Build GRCh38
rs913254603 Chr.19: 19258425-19258433 on Build GRCh38
rs762258885 Chr.19: 19258518-19258518 on Build GRCh38
rs774998208 Chr.19: 19258506-19258506 on Build GRCh38
rs954355242 Chr.19: 19258525-19258525 on Build GRCh38
rs940166115 Chr.19: 19258389-19258389 on Build GRCh38
rs1010479933 Chr.19: 19258473-19258473 on Build GRCh38
rs907120008 Chr.19: 19258446-19258446 on Build GRCh38
rs770880343 Chr.19: 19258474-19258474 on Build GRCh38
rs753157757 Chr.19: 19258447-19258447 on Build GRCh38
rs985629253 Chr.19: 19258364-19258364 on Build GRCh38
rs745516424 Chr.19: 19258478-19258478 on Build GRCh38
rs145531491 Chr.19: 19258442-19258442 on Build GRCh38
rs368786976 Chr.19: 19258512-19258512 on Build GRCh38
rs891910642 Chr.19: 19258469-19258469 on Build GRCh38
rs1005952177 Chr.19: 19258465-19258465 on Build GRCh38
rs751565044 Chr.19: 19258565-19258565 on Build GRCh38
rs769419171 Chr.19: 19258495-19258495 on Build GRCh38
rs762575269 Chr.19: 19258509-19258509 on Build GRCh38
rs564862847 Chr.19: 19258539-19258539 on Build GRCh38
rs781080608 Chr.19: 19258477-19258477 on Build GRCh38
rs112339374 Chr.19: 19258460-19258460 on Build GRCh38
rs931621324 Chr.19: 19258421-19258421 on Build GRCh38

View Full Product Details