Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 79362522-79362747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITM2A
Entrez Gene ID
9452
Gene Name
integral membrane protein 2A
Gene Aliases
BRICD2A, E25A
UniGene
Hs.17109
Gene Chromosome Location
Chr.X: 79360384-79367552 on Build GRCh38
Transcripts
NM_001171581.1 NM_004867.4

SNP Detail

SNP ID Chromosome Location
rs755313086 Chr.X: 79362634-79362634 on Build GRCh38
rs762769122 Chr.X: 79362551-79362551 on Build GRCh38
rs756550098 Chr.X: 79362601-79362601 on Build GRCh38
rs751719372 Chr.X: 79362555-79362555 on Build GRCh38
rs780824997 Chr.X: 79362658-79362658 on Build GRCh38
rs371317824 Chr.X: 79362565-79362565 on Build GRCh38
rs997563108 Chr.X: 79362604-79362604 on Build GRCh38
rs780574393 Chr.X: 79362609-79362609 on Build GRCh38
rs1036820002 Chr.X: 79362624-79362624 on Build GRCh38
rs770280828 Chr.X: 79362706-79362706 on Build GRCh38
rs770767743 Chr.X: 79362646-79362646 on Build GRCh38
rs369176321 Chr.X: 79362716-79362716 on Build GRCh38
rs1751134 Chr.X: 79362553-79362553 on Build GRCh38
rs754328122 Chr.X: 79362620-79362620 on Build GRCh38
rs368755402 Chr.X: 79362562-79362562 on Build GRCh38
rs775445464 Chr.X: 79362687-79362687 on Build GRCh38
rs745421086 Chr.X: 79362660-79362660 on Build GRCh38
rs746656914 Chr.X: 79362642-79362642 on Build GRCh38
rs776166161 Chr.X: 79362705-79362706 on Build GRCh38
rs778502438 Chr.X: 79362686-79362686 on Build GRCh38
rs762811211 Chr.X: 79362695-79362695 on Build GRCh38
rs762006936 Chr.X: 79362711-79362711 on Build GRCh38
rs750553735 Chr.X: 79362582-79362582 on Build GRCh38
rs746220951 Chr.X: 79362553-79362554 on Build GRCh38
rs184180205 Chr.X: 79362701-79362701 on Build GRCh38
rs149783799 Chr.X: 79362665-79362665 on Build GRCh38
rs901968300 Chr.X: 79362574-79362574 on Build GRCh38
rs977386253 Chr.X: 79362671-79362671 on Build GRCh38
rs779425778 Chr.X: 79362641-79362641 on Build GRCh38
rs1013517005 Chr.X: 79362668-79362668 on Build GRCh38
rs781612566 Chr.X: 79362552-79362553 on Build GRCh38
rs768371946 Chr.X: 79362699-79362699 on Build GRCh38
rs35236366 Chr.X: 79362662-79362663 on Build GRCh38

View Full Product Details