Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33338578-33338807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK3
Entrez Gene ID
10114
Gene Name
homeodomain interacting protein kinase 3
Gene Aliases
DYRK6, FIST3, PKY, YAK1
UniGene
Hs.201918
Gene Chromosome Location
Chr.11: 33256672-33357023 on Build GRCh38
Transcripts
NM_001048200.2 NM_001278162.1 NM_001278163.1 NM_005734.4 XM_005252729.2 XM_017017076.1 XM_017017077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415621 c.1384G>A p.E462K Chr.11: 33338799-33338799 on Build GRCh38
4647311 c.1346T>C p.L449S Chr.11: 33338761-33338761 on Build GRCh38
6583069 c.1344A>G p.T448T Chr.11: 33338759-33338759 on Build GRCh38
6347107 c.1387G>T p.A463S Chr.11: 33338802-33338802 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375652655 Chr.11: 33338745-33338745 on Build GRCh38
rs1057047349 Chr.11: 33338658-33338658 on Build GRCh38
rs952539754 Chr.11: 33338705-33338705 on Build GRCh38
rs772847434 Chr.11: 33338599-33338600 on Build GRCh38
rs750942460 Chr.11: 33338751-33338752 on Build GRCh38
rs201870098 Chr.11: 33338707-33338707 on Build GRCh38
rs753374157 Chr.11: 33338729-33338730 on Build GRCh38
rs375645561 Chr.11: 33338604-33338604 on Build GRCh38
rs895686633 Chr.11: 33338659-33338659 on Build GRCh38
rs541593457 Chr.11: 33338681-33338681 on Build GRCh38
rs765863086 Chr.11: 33338711-33338713 on Build GRCh38
rs763075074 Chr.11: 33338736-33338736 on Build GRCh38
rs766663471 Chr.11: 33338737-33338737 on Build GRCh38
rs778423567 Chr.11: 33338697-33338697 on Build GRCh38
rs74824265 Chr.11: 33338690-33338690 on Build GRCh38
rs959219326 Chr.11: 33338770-33338770 on Build GRCh38
rs761926809 Chr.11: 33338716-33338716 on Build GRCh38
rs572871919 Chr.11: 33338660-33338660 on Build GRCh38
rs544468970 Chr.11: 33338744-33338744 on Build GRCh38
rs1012845655 Chr.11: 33338666-33338666 on Build GRCh38
rs185305513 Chr.11: 33338657-33338657 on Build GRCh38
rs754433359 Chr.11: 33338611-33338611 on Build GRCh38
rs558033790 Chr.11: 33338599-33338599 on Build GRCh38
rs372662681 Chr.11: 33338600-33338600 on Build GRCh38
rs753203801 Chr.11: 33338768-33338768 on Build GRCh38
rs765412633 Chr.11: 33338719-33338719 on Build GRCh38
rs575196538 Chr.11: 33338726-33338726 on Build GRCh38
rs999297828 Chr.11: 33338617-33338617 on Build GRCh38
rs369717340 Chr.11: 33338603-33338604 on Build GRCh38
rs143681221 Chr.11: 33338612-33338612 on Build GRCh38
rs781620516 Chr.11: 33338753-33338753 on Build GRCh38
rs1034640054 Chr.11: 33338765-33338765 on Build GRCh38
rs1015378673 Chr.11: 33338723-33338723 on Build GRCh38

View Full Product Details