Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156495913-156496174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD2
Entrez Gene ID
2820
Gene Name
glycerol-3-phosphate dehydrogenase 2
Gene Aliases
GDH2, GPDM, mGPDH
UniGene
Hs.512382
Gene Chromosome Location
Chr.2: 156435290-156586403 on Build GRCh38
Transcripts
NM_000408.4 NM_001083112.2 XM_005246469.2 XM_011510977.2 XM_011510978.2 XM_017003830.1 XM_017003831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1325850 c.111G>A p.L37L Chr.2: 156496052-156496052 on Build GRCh38
6373476 c.154A>G p.R52G Chr.2: 156496095-156496095 on Build GRCh38
7154815 c.203C>G p.S68C Chr.2: 156496144-156496144 on Build GRCh38
7289529 c.152A>T p.N51I Chr.2: 156496093-156496093 on Build GRCh38
6364557 c.191T>C p.L64S Chr.2: 156496132-156496132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200523528 Chr.2: 156496078-156496078 on Build GRCh38
rs1002625732 Chr.2: 156495978-156495978 on Build GRCh38
rs917904916 Chr.2: 156495976-156495976 on Build GRCh38
rs903017056 Chr.2: 156495958-156495958 on Build GRCh38
rs371555200 Chr.2: 156496057-156496057 on Build GRCh38
rs755453568 Chr.2: 156496010-156496010 on Build GRCh38
rs764884977 Chr.2: 156496062-156496062 on Build GRCh38
rs952411607 Chr.2: 156496054-156496054 on Build GRCh38
rs145159302 Chr.2: 156496136-156496136 on Build GRCh38
rs374156040 Chr.2: 156496044-156496044 on Build GRCh38
rs77492341 Chr.2: 156496142-156496142 on Build GRCh38
rs761390657 Chr.2: 156496059-156496059 on Build GRCh38
rs760580252 Chr.2: 156496051-156496051 on Build GRCh38
rs765791937 Chr.2: 156496121-156496121 on Build GRCh38
rs376686130 Chr.2: 156496055-156496055 on Build GRCh38
rs781674686 Chr.2: 156496016-156496016 on Build GRCh38
rs202032384 Chr.2: 156496028-156496028 on Build GRCh38
rs771911363 Chr.2: 156496046-156496046 on Build GRCh38
rs770019110 Chr.2: 156496023-156496023 on Build GRCh38
rs749906240 Chr.2: 156496090-156496090 on Build GRCh38
rs1035416961 Chr.2: 156495981-156495981 on Build GRCh38
rs543520570 Chr.2: 156496073-156496073 on Build GRCh38
rs376816166 Chr.2: 156496026-156496026 on Build GRCh38
rs558415343 Chr.2: 156495977-156495977 on Build GRCh38
rs747513119 Chr.2: 156496000-156496000 on Build GRCh38
rs757992246 Chr.2: 156496104-156496104 on Build GRCh38
rs748309922 Chr.2: 156496039-156496039 on Build GRCh38
rs748420947 Chr.2: 156496018-156496018 on Build GRCh38
rs373389772 Chr.2: 156496066-156496066 on Build GRCh38
rs749484773 Chr.2: 156496127-156496127 on Build GRCh38
rs949316593 Chr.2: 156496002-156496002 on Build GRCh38
rs576961259 Chr.2: 156496050-156496050 on Build GRCh38

View Full Product Details